Kinesis® TELOS® HPLC Columns, 150 mm L
Chromatography columns in a comprehensive array of high-density bonded phases
  • Supported by 1-cm guard cartridges with identical packing material Includes test chromatogram
TELOS columns deliver better performance, reliability, and reproducibility, and are available in a comprehensive range of high-density bonded phases. Columns come packed with a variety of standard and wide pore materials. Each is individually tested for quality and includes a test chromatogram, which lists the test conditions and analytes.
$653.00 - $3,908.50USD / Each
42 variations of this product are available.
See All Product Options Below
42 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Particle Size (µm) Phase
Availability Pricing
COMPARE
2.1 3 C18
$817.50USD / Each
COMPARE
4.6 5 PFP
$758.50USD / Each
COMPARE
10 10 C18
$1,591.00USD / Each
COMPARE
3.9 5 C18
$758.50USD / Each
COMPARE
4.6 3 Phenyl hexyl
$898.50USD / Each
COMPARE
2.1 5 MDC-C8
$781.00USD / Each
COMPARE
4.6 5 ZE-EX-C18
$766.00USD / Each
COMPARE
4.6 5 MDC-C18
$781.00USD / Each
COMPARE
19 5 C18
$3,908.50USD / Each
COMPARE
4.6 3 Phenyl
$817.50USD / Each
COMPARE
4.6 5 Phenyl
$758.50USD / Each
COMPARE
2.1 5 MSP-C4
$758.50USD / Each
COMPARE
4.6 5 Phenyl hexyl
$825.00USD / Each
COMPARE
4.6 5 MSP-C4
$758.50USD / Each
COMPARE
4.6 5 ZE-E-C8
$758.50USD / Each
COMPARE
3 5 MDC-C18
$781.00USD / Each
COMPARE
2.1 1.8 C18 Polar
$1,171.00USD / Each
COMPARE
4 5 MSP-C4
$758.50USD / Each
COMPARE
4.6 3 C18 Polar
$937.00USD / Each
COMPARE
4.6 5 C8
$758.50USD / Each
Showing 1 - 20 of 42