Kinesis® TELOS® HPLC Columns, 250 mm L
Chromatography columns in a comprehensive array of high-density bonded phases
  • Supported by 1-cm guard cartridges with identical packing material Includes test chromatogram
TELOS columns deliver better performance, reliability, and reproducibility, and are available in a comprehensive range of high-density bonded phases. Columns come packed with a variety of standard and wide pore materials. Each is individually tested for quality and includes a test chromatogram, which lists the test conditions and analytes.
$744.00 - $3,134.50USD / Each
24 variations of this product are available.
See All Product Options Below
24 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Particle Size (µm) Phase
Availability Pricing
COMPARE
4.6 5 Phenyl
$817.50USD / Each
COMPARE
4 5 Aminohexadecyl
$744.00USD / Each
COMPARE
10 5 C18 AB-D
$2,202.00USD / Each
COMPARE
4.6 5 MSP-C4
$817.50USD / Each
COMPARE
4 5 MSP-C4
$817.50USD / Each
COMPARE
10 5 Silica-2
$1,364.50USD / Each
COMPARE
21.2 5 Silica-2
$3,134.50USD / Each
COMPARE
4 5 MDC-C18
$839.50USD / Each
COMPARE
2.1 5 MDC-C18
$839.50USD / Each
COMPARE
4.6 10 C18
$758.50USD / Each
COMPARE
3 5 MSP-C4
$817.50USD / Each
COMPARE
4.6 5 C18 Polar
$817.50USD / Each
COMPARE
4.6 5 Z-RX-C8
$825.00USD / Each
COMPARE
2.1 5 MSP-C4
$817.50USD / Each
COMPARE
4.6 5 Phenyl hexyl
$898.50USD / Each
COMPARE
4.6 5 Cyano
$1,238.00USD / Each
COMPARE
3 5 MDC-C18
$839.50USD / Each
COMPARE
4.6 5 MDC-C18
$839.50USD / Each
COMPARE
4.6 5 C18
$817.50USD / Each
COMPARE
4.6 5 PFP
$817.50USD / Each
Showing 1 - 20 of 24