Kodiak® Level Switches, 316 Stainless Steel
Accurate and rugged level monitoring at higher temperatures
  • Constructed of durable 316 stainless steel (SS)
These all-stainless-steel units are exceptionally accurate and rugged for level monitoring at higher temperatures...to 300°F (GM424113)... and in pressurized or corrosive liquids. Widely used in chemical plants and systems. For use in oils, water, chemicals.
$117.60 - $131.60USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Pressure (PSI)
Availability Pricing
Kodiak 1950 316 SS Level Switch, SPST, 0.75 SG, 1/4"NPT(M); 20VA EW-68964-27 Mfr # 1950
COMPARE
750
$117.60USD / Each
Kodiak 26717 316 SS Level Switch, SPST, 0.75 SG, 1/4"NPT(M); 100VA EW-68964-29 Mfr # 26717
COMPARE
750
$131.60USD / Each