L-K Industries Benchmark Heated Oil Centrifuges
Safety first—Class 1, Division 2 certified models available
  • Programmable digital controls maintain sample temperature and RPM accurately
  • Motor controller-integrated timer ensures test method compliance
  • Integral mounting flanges allow secure attachment to benchtop, stand, or table
  • Conformance: ASTM D91, D96, D893, D1796, D2273, D2709, and D4007
  • Shipping weight: 110 lb (50 kg)
These benchtop laboratory centrifuges help you meet the requirements of a variety of ASTM methods for oil testing. Program temperature and speed with digital precision, set motor control-integrated timer for test duration, and start the run. Simple controls save time, automated functions allow you to perform other tasks, and safety features protect the lab.
Choose explosion-proof models for hazardous location operation. Additional safety features include a lid-locking system preventing motor operation when open, emergency power-off palm button, and indicator lamps.
Order sample tubes separately.
Note: All models must be hard-wired by a qualified electrician with 12 AWG wire.
$10,656.00 - $18,240.00USD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Electrical Plug Type Power (VAC) Rotor Type
Availability Pricing
L-K Industries Class 1 Div 2 Heated Oil Centrifuge for Long Cone Tubes, 120 VAC EW-17700-00 Mfr # LB14-C120C
COMPARE
None, hard-wiring required, Class 1 Div 2 approved 120 Long cone tube type
$18,240.00USD / Each
L-K Industries Class 1 Div 2 Heated Oil Centrifuge for Short Cone Tubes, 120 VAC EW-17700-01 Mfr # LB14-D120C
COMPARE
None, hard-wiring required, Class 1 Div 2 approved 120 Short cone tube type
$18,240.00USD / Each
L-K Industries Class 1 Div 2 Heated Oil Centrifuge for Trace Sediment Tubes, 120 VAC EW-17700-02 Mfr # LB14-E120C
COMPARE
None, hard-wiring required, Class 1 Div 2 approved 120 Trace sediment tube type
$18,240.00USD / Each
L-K Industries Class 1 Div 2 Heated Oil Centrifuge for Long Cone Tubes, 220 VAC EW-17700-03 Mfr # LB14-C220C
COMPARE
None, hard-wiring required, Class 1 Div 2 approved 220 Long cone tube type
$18,240.00USD / Each
L-K Industries Class 1 Div 2 Heated Oil Centrifuge for Short Cone Tubes, 220 VAC EW-17700-04 Mfr # LB14-D220C
COMPARE
None, hard-wiring required, Class 1 Div 2 approved 220 Short cone tube type
$18,240.00USD / Each
L-K Industries Class 1 Div 2 Heated Oil Centrifuge for Trace Sediment Tubes, 220 VAC EW-17700-05 Mfr # LB14-E220C
COMPARE
None, hard-wiring required, Class 1 Div 2 approved 220 Trace sediment tube type
$18,240.00USD / Each
L-K Industries Heated Oil Centrifuge for Long Cone Tubes, 120 VAC EW-17700-06 Mfr # LBS-C120
COMPARE
None, hard-wiring required 120 Long cone tube type
$10,760.00USD / Each
L-K Industries Heated Oil Centrifuge for Short Cone Tubes, 120 VAC EW-17700-07 Mfr # LBS-D120
COMPARE
None, hard-wiring required 120 Short cone tube type
$10,656.00USD / Each
L-K Industries Heated Oil Centrifuge for Trace Sediment Tubes, 120 VAC EW-17700-08 Mfr # LBS-E120
COMPARE
None, hard-wiring required 120 Trace sediment tube type
$10,656.00USD / Each
L-K Industries Heated Oil Centrifuge for Long Cone Tubes, 220 VAC EW-17700-09 Mfr # LBS-C220
COMPARE
None, hard-wiring required 220 Long cone tube type
$10,656.00USD / Each
L-K Industries Heated Oil Centrifuge for Short Cone Tubes, 220 VAC EW-17700-10 Mfr # LBS-D220
COMPARE
None, hard-wiring required 220 Short cone tube type
$10,656.00USD / Each
L-K Industries Heated Oil Centrifuge for Trace Sediment Tubes, 220 VAC EW-17700-11 Mfr # LBS-E220
COMPARE
None, hard-wiring required 220 Trace sediment tube type
$10,656.00USD / Each
Showing 1- 12 of 12