L-K Industries Centrifuge Tubes
Ideal for use in regulatory environments
  • Unique serial number on each tube provides precise traceability
  • Permanently etched graduated marks improve visibility
  • Sand spot simplifies marking tubes in the field and lab
L-K Industries oil centrifuge tubes come configured with either a cork- or rubber-tooled top, or a wide- or standard-thread top. The tubes feature 2-mm thick wall configurations offering durability and high RCF capability.
The glass tubes come with standard or detailed permanently etched graduated markings. Tubes with detailed markings are ideal for test sites or wells that produce more water than what is typical.
$27.25 - $199.00USD / Each
121 variations of this product are available.
See All Product Options Below
121 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
L-K Industries Centrifuge Trace Sediment Tube, Tooled, Detailed mL, verified, 1/Ea EW-17700-60 Mfr # S12EFV15
COMPARE
Centrifuge Trace Sediment Tube, Tooled, Detailed mL, verified, 1/Ea
$127.00USD / Each
L-K Industries Centrifuge Trace Sediment Tube, Tooled, Detailed %, certified, 1/Ea EW-17700-64 Mfr # S12E2AV15
COMPARE
Centrifuge Trace Sediment Tube, Tooled, Detailed %, certified, 1/Ea
$169.00USD / Each
L-K Industries Centrifuge Trace Sediment Tube, Threaded, mL Markings, Certified, 1/Ea EW-17700-67 Mfr # S10ETHDAV15
COMPARE
Centrifuge Trace Sediment Tube, Threaded, mL Markings, Certified, 1/Ea
$167.00USD / Each
L-K Industries Centrifuge Trace Sediment Tube, Wide, % Markings, Verified, 1/Ea EW-17900-08 Mfr # S10E2WTHDFV15
COMPARE
Centrifuge Trace Sediment Tube, Wide, % Markings, Verified, 1/Ea
$164.00USD / Each
L-K Industries Centrifuge Trace Sediment Tube, Wide, Detailed mL, Verified, 1/Ea EW-17900-11 Mfr # S12EWTHDFV15
COMPARE
Centrifuge Trace Sediment Tube, Wide, Detailed mL, Verified, 1/Ea
$169.00USD / Each
L-K Industries Centrifuge Trace Sediment Tube, Wide, Detailed %, 1/Ea EW-17900-13 Mfr # S12E2WTHD
COMPARE
Centrifuge Trace Sediment Tube, Wide, Detailed %, 1/Ea
$160.00USD / Each
L-K Industries Centrifuge Pear Shaped Tube, Tooled, mL Markings, 1/Ea EW-17900-16 Mfr # S10B
COMPARE
Centrifuge Pear Shaped Tube, Tooled, mL Markings, 1/Ea
$96.00USD / Each
L-K Industries Centrifuge Pear Shaped Tube, Tooled, % Markings, 1/Ea EW-17900-19 Mfr # S10B2
COMPARE
Centrifuge Pear Shaped Tube, Tooled, % Markings, 1/Ea
$95.50USD / Each
L-K Industries Centrifuge Pear Shaped Tube, Tooled, % Markings, Certified, 1/Ea EW-17900-21 Mfr # S10B2AV15
COMPARE
Centrifuge Pear Shaped Tube, Tooled, % Markings, Certified, 1/Ea
$160.00USD / Each
L-K Industries Centrifuge Pear Shaped Tube, Tooled, Detailed mL, Certified, 1/Ea EW-17900-24 Mfr # S12EAV15
COMPARE
Centrifuge Pear Shaped Tube, Tooled, Detailed mL, Certified, 1/Ea
$147.00USD / Each
L-K Industries Centrifuge Long Cone Tube, Threaded, mL Markings, Verified, 1/Ea EW-17900-41 Mfr # S10CTHDFV15
COMPARE
Centrifuge Long Cone Tube, Threaded, mL Markings, Verified, 1/Ea
$94.00USD / Each
L-K Industries Centrifuge Long Cone Tube, Threaded, Detailed %, 1/Ea EW-17900-50 Mfr # S12C2THDFV15
COMPARE
Centrifuge Long Cone Tube, Threaded, Detailed %, 1/Ea
$120.00USD / Each
L-K Industries Centrifuge Long Cone Tube, Threaded, Detailed %, Certified, 1/Ea EW-17900-52 Mfr # S10CWTHD
COMPARE
Centrifuge Long Cone Tube, Threaded, Detailed %, Certified, 1/Ea
$124.00USD / Each
L-K Industries Centrifuge Long Cone Tube, Wide, Detailed %, Verified, 1/Ea EW-17900-63 Mfr # S12C2WTHDAV15
COMPARE
Centrifuge Long Cone Tube, Wide, Detailed %, Verified, 1/Ea
$175.00USD / Each
L-K Industries Centrifuge Short Cone Tube, Tooled, % Markings, Verified, 1/Ea EW-17900-69 Mfr # S10D2AV15
COMPARE
Centrifuge Short Cone Tube, Tooled, % Markings, Verified, 1/Ea
$126.00USD / Each
L-K Industries Centrifuge Short Cone Tube, Tooled, % Markings, Certified, 1/Ea EW-17900-70 Mfr # S12D
COMPARE
Centrifuge Short Cone Tube, Tooled, % Markings, Certified, 1/Ea
$92.50USD / Each
L-K Industries Centrifuge Short Cone Tube, Threaded, Detailed %, 1/Ea EW-17900-85 Mfr # S12D2THD
COMPARE
Centrifuge Short Cone Tube, Threaded, Detailed %, 1/Ea
$115.00USD / Each
L-K Industries Centrifuge Short Cone Tube, Wide, % Markings, Verified, 1/Ea EW-17900-92 Mfr # S10D2WTHDFV15
COMPARE
Centrifuge Short Cone Tube, Wide, % Markings, Verified, 1/Ea
$109.00USD / Each
L-K Industries Centrifuge Short Cone Tube, Wide, Detailed mL, Verified, 1/Ea EW-17900-95 Mfr # S12DWTHDFV15
COMPARE
Centrifuge Short Cone Tube, Wide, Detailed mL, Verified, 1/Ea
$130.00USD / Each
L-K Industries Centrifuge Short Cone Tube, Wide, Detailed mL, Certified, 1/Ea EW-17900-96 Mfr # S12DWTHDAV15
COMPARE
Centrifuge Short Cone Tube, Wide, Detailed mL, Certified, 1/Ea
$165.00USD / Each
Showing 1 - 20 of 121