L-K Industries Criterion Water Bath
Preheat samples for faster, easier testing
Criterion Water Baths are Class 1 Division 2 certified for use in hazardous locations. Programmable set point and digital temperature display ensure repeatable results. Included bath cover minimizes evaporation. Choose models with space for two one-gallon solvent bottles, fourteen long-cone tubes, or twelve short-cone tubes.
Note: all units must be hard-wired by a qualified electrician.
$5,156.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Description
Availability Pricing
L-K Industries Criterion Water Bath, two 1 gal bottles, 120 VAC EW-17700-20 Mfr # WB-T2-120
COMPARE
120 Water Bath, two 1 gal bottles, 120 VAC
$5,156.00USD / Each
L-K Industries Criterion Water Bath, two 1 gal bottles, 240 VAC EW-17700-21 Mfr # WB-T2-240
COMPARE
240 Water Bath, two 1 gal bottles, 240 VAC
$5,156.00USD / Each
L-K Industries Criterion Water Bath, long cone or trace sediment tubes, 120 VAC EW-17700-22 Mfr # WB-T-14C-120
COMPARE
120 Water Bath, long cone or trace sediment tubes, 120 VAC
$5,156.00USD / Each
L-K Industries Criterion Water Bath, long cone or trace sediment tubes, 240 VAC EW-17700-23 Mfr # WB-T-14C-240
COMPARE
240 Water Bath, long cone or trace sediment tubes, 240 VAC
$5,156.00USD / Each
L-K Industries Criterion Water Bath, short cone tubes, 120 VAC EW-17700-24 Mfr # WB-T-12D-120
COMPARE
120 Water Bath, short cone tubes, 120 VAC
$5,156.00USD / Each
L-K Industries Criterion Water Bath, short cone tubes, 240 VAC EW-17700-25 Mfr # WB-T-12D-240
COMPARE
240 Water Bath, short cone tubes, 240 VAC
$5,156.00USD / Each