L-K Industries Melton Heated Oil Centrifuges
Versatile—each cup is individually heated and controlled
  • Separate heating elements and thermostats allow different temperature conditions at the same time
  • Built-in mechanical tachometer shows current rotational speed at a glance
  • Hand brake brings rotor to a stop
This benchtop Melton-type oil centrifuge is factory preset to keep samples heated at 150°F (66°C), but the temperature can be adjusted by simply turning an adjustment screw. Built-in mechanical tachometer and hand brake keep operation simple.
Order sample tubes and companion sample heater separately.
Note: All models must be hard-wired by a qualified electrician.
$10,000.00 - $10,010.00USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Tube Capacity
Availability Pricing
L-K Industries Melton Oil Centrifuge for Pear Shaped Tubes, 115 VAC EW-17700-12 Mfr # B-4-115-C
COMPARE
120 100-mL Pear shape
$10,000.00USD / Each
L-K Industries Melton Oil Centrifuge for Pear Shaped Tubes, 220 VAC EW-17700-13 Mfr # B-4-220-C
COMPARE
220 100-mL Pear shape
$10,000.00USD / Each
L-K Industries Melton Oil Centrifuge for Long Cone Tubes, 115 VAC EW-17700-14 Mfr # C-4-115-C
COMPARE
120 100-mL Long cone
$10,000.00USD / Each
L-K Industries Melton Oil Centrifuge for Long Cone Tubes, 220 VAC EW-17700-15 Mfr # C-4-220-C
COMPARE
220 100-mL Long cone
$10,010.00USD / Each
L-K Industries Melton Oil Centrifuge for Short Cone Tubes, 115 VAC EW-17700-16 Mfr # D-4-115-C
COMPARE
120 100-mL Short cone
$10,000.00USD / Each
L-K Industries Melton Oil Centrifuge for Short Cone Tubes, 220 VAC EW-17700-17 Mfr # D-4-220-C
COMPARE
220 100-mL Short cone
$10,000.00USD / Each
L-K Industries Melton Oil Centrifuge for Trace Sediment Tubes, 115 VAC EW-17700-18 Mfr # E-4-115-C
COMPARE
120 100-mL Trace sediment
$10,000.00USD / Each
L-K Industries Melton Oil Centrifuge for Trace Sediment Tubes, 220 VAC EW-17700-19 Mfr # E-4-220-C
COMPARE
220 100-mL Trace sediment
$10,000.00USD / Each
Showing 1- 8 of 8