L-K Industries Transport Series Sample Heaters
Preheat samples for easier testing
Transport Series Sample Heaters run on the 12 VDC supply created by a running vehicle. Compact and lightweight design with flip-up lid make travel with this heater no hassle at all. Simple high/off/low switch operation with quick sample heat-up time minimize field testing time. Rugged steel construction withstands rigors of field use.
$596.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Block Number Of Openings Block - Tube / Vial Size Accepted (mL) Description
Availability Pricing
L-K Industries Transport Series Portable Sample Heater, short cone tubes, 12 VDC EW-17700-36 Mfr # D212H
COMPARE
2 100 Portable Sample Heater, short cone tubes, 12 VDC
$596.00USD / Each
L-K Industries Transport Series Portable Sample Heater, finger tubes, 12 VDC EW-17700-37 Mfr # A412H
COMPARE
4 12.5 Portable Sample Heater, finger tubes, 12 VDC
$596.00USD / Each