L-K Industries Transportable Centrifuge
Get fast test results on crude oil haulers
  • Rugged design offers longer life and less downtime
  • Compact design makes installation and maintenance easy
These transportable centrifuges are ideal for determining sediment and water content on crude oil haulers. The centrifuges are available in analog and digital models with a heated or nonheated configuration, and either a 2- or 4-place Trunnion arm.
Each model features a sturdy outer steel case with reinforced mounting rails for outdoor use and variable-speed motor controller with epoxy-coated electronics.
Order sample tubes and companion sample heater separately.
$1,564.00 - $4,396.00USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Speed Control Tube Capacity
Availability Pricing
L-K Industries Transportable Heated Digital Centrifuge, Two Short Cone Tubes, 12 V EW-17700-42 Mfr # TRANS-9100
COMPARE
Digital 2 x 100 mL short cone
$3,182.00USD / Each
L-K Industries Heated Digital Centrifuge, Short Cone Tubes, 115 VAC EW-17700-43 Mfr # TRANS-9100/115
COMPARE
Digital 2 x 100 mL short cone
$3,962.00USD / Each
L-K Industries Transportable Heated Analog Centrifuge, Two Short Cone Tubes, 12 VDC EW-17700-44 Mfr # TRANS-7100
COMPARE
121 Analog 2 x 100 mL short cone
$2,392.00USD / Each
L-K Industries Transportable Heated Analog Centrifuge, Four Short Cone Tubes, 12 VDC EW-17700-45 Mfr # TRANS-7100/4
COMPARE
121 Analog 4 x100 mL short cone
$3,098.00USD / Each
L-K Industries Transportable Heated Digital Centrifuge, Four Short Cone Tubes, 12 V EW-17700-46 Mfr # TRANS-9100/4
COMPARE
Digital 4 x100 mL short cone
$3,732.00USD / Each
L-K Industries Heated Digital Centrifuge, Four Short Cone Tubes, 115 VAC EW-17700-47 Mfr # TRANS-9100/4/115
COMPARE
Digital 4 x100 mL short cone
$4,282.00USD / Each
L-K Industries Heated Digital Centrifuge, 12 Finger Tubes, 115 VAC EW-17700-48 Mfr # TRANS-9100/ A12/115
COMPARE
Digital 12 x 12.5 mL finger
$4,396.00USD / Each
L-K Industries Transportable Analog Centrifuge, Two Pear Shaped Tubes, 12 VDC EW-17700-49 Mfr # TRANS-12/B2
COMPARE
Analog 2 x 100 mL pear shaped
$1,716.00USD / Each
L-K Industries Transportable Analog Centrifuge, Four Pear Shaped Tubes, 12 VDC EW-17700-50 Mfr # TRANS-12/B4
COMPARE
Analog 4 x 100 mL pear shaped
$1,924.00USD / Each
L-K Industries Transportable Analog Centrifuge, Two Short Cone Tubes, 12 VDC EW-17700-51 Mfr # TRANS-12/D2
COMPARE
Analog 2 x 100 mL short cone
$1,564.00USD / Each
Showing 1- 10 of 10