Lab Companion Vacuum Ovens
Feature wide temperature range for a variety of test applications
  • Perform a variety of applications from moisture determination, aging test, and sterilization to solvent separation under controlled atmosphere conditions
  • Ideal for separating solvents from a solution without high-temperature drying
  • Heat-tempered full-view window allows continuous monitoring of chamber
  • User-friendly digital control panel simplifies operation
Separate vacuum and vent ports allow greater control of vacuum rates. Vacuum levels are maintained by a tightly sealed door gasket and push-button latch. Block heaters and anodized aluminum plate shelves provide uniform heating throughout the oven chamber.
$4,990.00 - $6,150.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (cu ft) Power (VAC) Description
Availability Pricing
Lab Companion LIKE NEW 52402-96
Steals and Deals
EW-00299-WK Mfr # 52402-96 LIKE NEW
COMPARE
2 120 Vacuum Oven 2.3 cu. ft.; 120VAC
Call For Price
Lab Companion AAH13116U Vacuum Oven 1.0 CU.FT; 120VAC EW-52402-95 Mfr # AAH13116U
COMPARE
1 120 Vacuum Oven 1.0 CU.FT; 120VAC
$4,990.00USD / Each
Lab Companion AAH13126U Vacuum Oven 2.3 cu. ft.; 120VAC EW-52402-96 Mfr # AAH13126U
COMPARE
2 120 Vacuum Oven 2.3 cu. ft.; 120VAC
$5,980.00USD / Each
Lab Companion AAH13115K Vacuum Oven 1.0 CU.FT; 230VAC EW-52402-97 Mfr # AAH13115K
COMPARE
1 230 Vacuum Oven 1.0 CU.FT; 230VAC
$5,430.00USD / Each
Lab Companion AAH13125K Vacuum Oven 2.3 cu. ft.; 230 VAC EW-52402-98 Mfr # AAH13125K
COMPARE
2 230 Vacuum Oven 2.3 cu. ft.; 230 VAC
$6,150.00USD / Each
Showing 1- 5 of 5
Need Help with this product?
Call Us At1-847-549-7600
Live chat