Lab-Crest Pressure Reaction Vessels
Accurately and safely observe your reactions!
  • Inert borosilicate glass construction with PVC coating for safety
  • Millimeter reference scale to mark changes in your sample volume
Lab-Crest® pressure reaction vessels allow for full observation of chemical reactions in a safe vessel. These vessels are made from heavy wall, inert borosilicate glass with a PVC safety coating. Each vessel is graduated with a millimeter reference scale. These pressure reaction vessels are sometimes referred to as “Fisher and Porter Bottles”. They are ideal for viewing reactions of mixed compounds and viewing temperature and pressure changes to liquids or gasses. Typical applications include hydrolysis, hydrogenation, polymerization, catalysis and isomerization. Other uses include corrosion studies, solubility studies and spray and pressure characteristics of compounds.
All reaction vessels are pressure tested to 2x their pressure rating. The vessels can be purchased in three standard sizes with various options.
Multi-Port Stirring Assembly includes 12-oz reaction vessel, rugged SS head and stirrer assembly for gathering chemical reaction data. This assembly provides a controlled environment for constant stirring, handles highly viscous materials, enables the use of pressure control gauges and temperature probes, and allows easy injection or extraction of samples.
Caution: Safety precaution should be used with glass under pressure. Maximum safe pressure depends on overall vessel condition. Additional shielding is strongly recommended.
$411.00 - $1,855.00USD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Capacity (oz) Material
Availability Pricing
Lab-Crest 110-452-0003 Pressure Reaction Vessel, Glass, 3 oz EW-63400-00 Mfr # 110-452-0003
COMPARE
88 3 Glass
$445.00USD / Each
Lab-Crest 110-452-0006 Pressure Reaction Vessel, Glass. 6 oz EW-63400-02 Mfr # 110-452-0006
COMPARE
170 6 Glass
$411.00USD / Each
Lab-Crest 110-452-0012 Pressure Reaction Vessel, Glass, 12 oz EW-63400-04 Mfr # 110-452-0012
COMPARE
355 12 Glass
$773.00USD / Each
Lab-Crest 110-106-0003 Pressure Reaction Vessel, with Coupling, Glass, 3 oz EW-63400-12 Mfr # 110-106-0003
COMPARE
88 3 Glass
$504.00USD / Each
Lab-Crest 110-106-0006 Pressure Reaction Vessel, with Coupling, Glass, 6 oz EW-63400-14 Mfr # 110-106-0006
COMPARE
170 6 Glass
$738.00USD / Each
Lab-Crest 110-106-0012 Pressure Reaction Vessel, with Coupling, Glass, 12 oz EW-63400-16 Mfr # 110-106-0012
COMPARE
355 12 Glass
$685.00USD / Each
Lab-Crest 110-207-0003 Reaction Vessel with Clear Plastic Shield and Coupling, 3 oz EW-63400-24 Mfr # 110-207-0003
COMPARE
88 3 Glass
$921.00USD / Each
Lab-Crest 110-207-0006 Reaction Vessel with Clear Plastic Shield and Coupling, 6 oz EW-63400-26 Mfr # 110-207-0006
COMPARE
170 6 Glass
$1,830.00USD / Each
Lab-Crest 110-207-0012 Reaction Vessel with Clear Plastic Shield and Coupling, 12 oz EW-63400-28 Mfr # 110-207-0012
COMPARE
355 12 Glass
$1,855.00USD / Each
Showing 1- 9 of 9