This product is now discontinued, View Related Products

LabChem Ammonium Hydroxide for Magnesium, 5%

Highest quality raw materials!
  • Reagents and solutions to meet your lab and research needs
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Call For Price

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Volume (mL) Description
Availability Pricing
LabChem Ammonium Hydroxide for Magnesium, 5% (v/v) (1+19); 500 mL EW-80009-17 Mfr # LC111901
COMPARE
500 Ammonium Hydroxide, 5% (v/v) (1+19), for Magnesium; 500 mL
Call For Price
LabChem Ammonium Hydroxide for Magnesium, 5% (v/v) (1+19); 4 L EW-80009-18 Mfr # LC111904
COMPARE
4000 Ammonium Hydroxide, 5% (v/v) (1+19), for Magnesium; 4 L
Call For Price
LabChem Ammonium Hydroxide for Magnesium, 5% (v/v) (1+19); 20 L EW-80009-19 Mfr # LC111905
COMPARE
20000 Ammonium Hydroxide, 5% (v/v) (1+19), for Magnesium; 20 L
Call For Price
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat