LabChem Chlorophenol Red, 0.04% Aqueous
Highest quality raw materials!
  • Reagents and solutions to meet your lab and research needs
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$52.50USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Volume (mL) Description
Availability Pricing
LabChem Chlorophenol Red, 0.04% Aqueous, pH 5.2-6.8 Yellow to Red; 1 L EW-80014-82 Mfr # LC130802
COMPARE
1000 Chlorophenol Red, 0.04% Aqueous, pH 5.2-6.8 Yellow to Red; 1 L
$52.50USD / Each