LabChem Methanol, 50% (v/v) (1+1)

Highest quality raw materials!
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$40.50 - $61.50USD / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Volume (mL) Description
Availability Pricing
LabChem Methanol, 50% (v/v) (1+1); 1 L EW-80028-86 Mfr # LC168302
COMPARE
1000 Methanol, 50% (v/v) (1 1); 1 L
$40.50USD / Each
LabChem Methanol, 50% (v/v) (1+1); 4 L EW-80028-87 Mfr # LC168304
COMPARE
4000 Methanol, 50% (v/v) (1 1); 4 L
$61.50USD / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat