LabChem Methyl Red, 0.02% in Methanol
Highest quality raw materials!
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$17.50 - $46.50USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Volume (mL) Description
Availability Pricing
LabChem Methyl Red, 0.02% in Methanol, pH 4.2-6.3 Pink to Yellow; 1 L EW-80029-17 Mfr # LC171002
COMPARE
1000 Methyl Red, 0.02% in Methanol, pH 4.2-6.3 Pink to Yellow; 1 L
$46.50USD / Each
LabChem Methyl Red, 0.02% in Methanol, pH 4.2-6.3 Pink to Yellow; 125 mL EW-80029-18 Mfr # LC171007
COMPARE
125 Methyl Red, 0.02% in Methanol, pH 4.2-6.3 Pink to Yellow; 125 mL
$17.50USD / Each