LabChem Methyl Red, 1.0% Aqueous
Highest quality raw materials!
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$18.50 - $48.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Volume (mL) Description
Availability Pricing
LabChem Methyl Red, 1.0% Aqueous, pH 4.2-6.3 Pink to Yellow; 500 mL EW-80029-22 Mfr # LC171401
COMPARE
500 Methyl Red, 1.0% Aqueous, pH 4.2-6.3 Pink to Yellow; 500 mL
$48.00USD / Each
LabChem Methyl Red, 1.0% Aqueous, pH 4.2-6.3 Pink to Yellow; 125 mL EW-80029-23 Mfr # LC171407
COMPARE
125 Methyl Red, 1.0% Aqueous, pH 4.2-6.3 Pink to Yellow; 125 mL
$18.50USD / Each