LabChem Nitrite (as Nitrogen) Standard, Preserved with Chloroform
Highest quality raw materials!
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$23.00 - $53.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Volume (mL) Description
Availability Pricing
LabChem Nitrite (as Nitrogen) Standard, 1000 ppm, with Chloroform (1 mL=1 mg N, 1 mL=3.28 mg NO₂ˉ ); 500 mL EW-80029-89 Mfr # LC179301
COMPARE
500 Nitrite (as Nitrogen) Standard, 1000 ppm, with Chloroform (1 mL=1 mg N, 1 mL=3.28 mg NO₂ˉ ); 500 mL
$53.00USD / Each
LabChem Nitrite (as Nitrogen) Standard, 1000 ppm, with Chloroform (1 mL=1 mg N, 1 mL=3.28 mg NO₂ˉ ); 125 mL EW-80029-90 Mfr # LC179307
COMPARE
125 Nitrite (as Nitrogen) Standard, 1000 ppm, with Chloroform (1 mL=1 mg N, 1 mL=3.28 mg NO₂ˉ ); 125 mL
$23.00USD / Each