LabChem o-Tolidine, 0.1%

Highest quality raw materials!
  • Reagents and solutions to meet your lab and research needs
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$45.30 - $52.20USD / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Volume (mL) Description
Availability Pricing
LabChem o-Tolidine, 0.1%; 500 mL EW-80054-57 Mfr # LC261601
COMPARE
500 o-Tolidine, 0.1%; 500 mL
$45.30USD / Each
LabChem o-Tolidine, 0.1%; 1 L EW-80054-58 Mfr # LC261602
COMPARE
1000 o-Tolidine, 0.1%; 1 L
$52.20USD / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat