LabChem Phenol, 5% (w/v)

Highest quality raw materials!
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$32.00 - $141.00USD / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Volume (mL) Description
Availability Pricing
LabChem Phenol, 5% (w/v); 500 mL EW-80030-52 Mfr # LC181951
COMPARE
500 Phenol, 5% (w/v); 500 mL
$32.00USD / Each
LabChem Phenol, 5% (w/v); 1 L EW-80030-53 Mfr # LC181952
COMPARE
1000 Phenol, 5% (w/v); 1 L
$45.50USD / Each
LabChem Phenol, 5% (w/v); 4 L EW-80030-54 Mfr # LC181954
COMPARE
4000 Phenol, 5% (w/v); 4 L
$141.00USD / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat