LabChem Phosphate (as Phosphorus) Standard
Highest quality raw materials!
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$23.50 - $48.50USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Concentration Volume (mL) Description
Availability Pricing
LabChem Phosphate (as Phosphorus) Standard, 1000 ppm (1 mL=1 mg P); 500 mL EW-80031-93 Mfr # LC185901
COMPARE
1000 ppm 500 Phosphate (as Phosphorus) Standard, 1000 ppm (1 mL=1 mg P); 500 mL
$48.50USD / Each
LabChem Phosphate (as Phosphorus) Standard, 50 ppm (1 mL=0.05 mg P); 500 mL EW-80031-94 Mfr # LC186001
COMPARE
50 ppm 500 Phosphate (as Phosphorus) Standard, 50 ppm (1 mL=0.05 mg P); 500 mL
$34.50USD / Each
LabChem Phosphate (as Phosphorus) Standard, 50 ppm (1 mL=0.05 mg P); 125 mL EW-80031-95 Mfr # LC186007
COMPARE
50 ppm 125 Phosphate (as Phosphorus) Standard, 50 ppm (1 mL=0.05 mg P); 125 mL
$23.50USD / Each