LabChem Potassium AA Standard
Highest quality raw materials!
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$19.00 - $39.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Concentration Volume (mL) Description
Availability Pricing
LabChem Potassium AA Standard, 1000 ppm (1 mL=1 mg K); 500 mL EW-80035-95 Mfr # LC201001
COMPARE
1000 ppm 500 Potassium AA Standard, 1000 ppm (1 mL=1 mg K); 500 mL
$39.00USD / Each
LabChem Potassium AA Standard, 1000 ppm (1 mL=1 mg K); 125 mL EW-80035-96 Mfr # LC201007
COMPARE
1000 ppm 125 Potassium AA Standard, 1000 ppm (1 mL=1 mg K); 125 mL
$19.00USD / Each
LabChem Potassium AA Standard, 100 ppm (1 mL=0.1 mg K); 500 mL EW-80035-97 Mfr # LC201101
COMPARE
100 ppm 500 Potassium AA Standard, 100 ppm (1 mL=0.1 mg K); 500 mL
$27.50USD / Each