LabChem Silver Nitrate, 0.171 N
Highest quality raw materials!
  • Reagents and solutions to meet your lab and research needs
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$296.00 - $2,968.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Volume (mL) Description
Availability Pricing
LabChem Silver Nitrate, 0.171 N (0.171 M) (1 mL=10 mg NaCl); 1 L EW-80037-25 Mfr # LC227202
COMPARE
1000 Silver Nitrate, 0.171 N (0.171 M) (1 mL=10 mg NaCl); 1 L
$296.00USD / Each
LabChem Silver Nitrate, 0.171 N (0.171 M) (1 mL=10 mg NaCl); 4 L EW-80037-26 Mfr # LC227204
COMPARE
4000 Silver Nitrate, 0.171 N (0.171 M) (1 mL=10 mg NaCl); 4 L
$921.50USD / Each
LabChem Silver Nitrate, 0.171 N (0.171 M) (1 mL=10 mg NaCl); 20 L EW-80037-27 Mfr # LC227205
COMPARE
20000 Silver Nitrate, 0.171 N (0.171 M) (1 mL=10 mg NaCl); 20 L
$2,968.00USD / Each