LabChem Thymolphthalein, 0.05% in Ethanol
Highest quality raw materials!
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$20.50 - $23.50USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Volume (mL) Description
Availability Pricing
LabChem Thymolphthalein, 0.05% in Ethanol, pH 9.3-10.5 Colorless to Blue; 500 mL EW-80054-42 Mfr # LC260701
COMPARE
500 Thymolphthalein, 0.05% in Ethanol, pH 9.3-10.5 Colorless to Blue; 500 mL
$23.50USD / Each
LabChem Thymolphthalein, 0.05% in Ethanol, pH 9.3-10.5 Colorless to Blue; 125 mL EW-80054-43 Mfr # LC260707
COMPARE
125 Thymolphthalein, 0.05% in Ethanol, pH 9.3-10.5 Colorless to Blue; 125 mL
$20.50USD / Each