Labconco Canopy Hoods
Vent non-toxic materials to keep your lab safe
  • Designed to be installed on a wall or suspended from the ceiling
Canopy Hoods are specifically designed to vent non-toxic materials such as heat, steam and odors from large or bulky apparatus that do not require a physical barrier such as ovens, steam baths and autoclaves. The hood may be installed on a wall or suspended from the ceiling for peninsular and corner locations. Coupled with a blower and ductwork (sold separately), the Canopy Hood can operate at a wide range of exhaust rates.
$3,525.00 - $3,985.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Interior Width (in) Power (VAC) Description
Availability Pricing
Labconco Canopy Hood, 72" EW-09072-64 Mfr # 7099000
COMPARE
72 115 Canopy Hood, 72"
$3,985.00USD / Each
Labconco Canopy Hood, 48" EW-09072-66 Mfr # 4799000
COMPARE
48 115 Fume hood canopy, 48"
$3,525.00USD / Each