Labconco Hooded Combo Kjeldahl Digestion/Distillation Apparatus
Ventilated hood captures excess heat and offers personnel safety
  • Easy-to-read temperature gauge
  • Remote-control flow valve allows cooling water volume adjustment
  • Counterflow heat exchanger controls back pressure, maximizes rate of distillation
  • Chemical-resistant CPVC fume manifold with heat-resistant PTFE nipples
The hooded combination Kjeldahl distillation and digestion apparatus safely vents excess heat from the laboratory. The hood includes three collars for connection to remotely located acid exhaust blower, hood heat exhaust blower, and auxiliary-air blower inlet. The models are constructed of epoxy-coated steel, reinforced with tubular steel and tempered safety glass, and horizontal-sliding sashes.
The lower digestion rack and upper distillation rack allows you to determine total nitrogen content within a variety of materials such as feeds, grains, soils, fertilizers, plant tissue, water, organic wastes, and food products. The distillation manifold is equipped with seamless stainless-steel tubes. No fittings or gaskets are required, eliminating potential leaks. To assure uniform results, the counterflow heat exchanger allows you to control back pressure and maximize the rate of distillation. The cooling water volume can be easily adjusted with the remote-control flow valve.
The connecting bulbs separates liquid from vapors prior to the vapors entering the condenser. It effectively traps caustic mist that may carry over to the recovery solution. The one-piece borosilicate glass delivery tube features a long neck, round bottom, and thick walls to help eliminate pressure fluctuations in the distillation phase. To provide evenly distributed heat and faster processing of samples, the curved ceramic electric heaters follow the round base of the Kjeldahl flask. Heaters accommodate 500- and 800-mL flasks (not included).
Three models are available with the capacity to digest and distill up to 6, 12 or 18 samples at a time.
$66,500.00 - $91,990.00USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flask Size (mL) Number of Positions Power (VAC)
Availability Pricing
Labconco 18-Place Hooded Combo Kjeldahl Digestion/Distillation Apparatus, Single Phase, 230 VAC, 60 Hz EW-78909-07 Mfr # 2117812
COMPARE
800 18 230, 1 phase
$91,990.00USD / Each
Labconco 18-Place Hooded Combo Kjeldahl Digestion/Distillation Apparatus, Three Phase; 230 VAC, 60 Hz EW-78909-08 Mfr # 2117813
COMPARE
800 18 230, 3 phase
$91,990.00USD / Each
Labconco 12-Place Hooded Combo Kjeldahl Digestion/Distillation Apparatus, Single Phase, 230 VAC, 60 Hz EW-78909-09 Mfr # 2117712
COMPARE
800 12 230, 1 phase
$81,470.00USD / Each
Labconco 12-Place Hooded Combo Kjeldahl Digestion/Distillation Apparatus, Three Phase; 230 VAC, 60 Hz EW-78909-10 Mfr # 2117713
COMPARE
800 12 230, 3 phase
$81,470.00USD / Each
Labconco 6-Place Hooded Combo Kjeldahl Digestion/Distillation Apparatus, Single Phase, 115 VAC, 60 Hz EW-78909-11 Mfr # 2117611
COMPARE
800 6 115, 1 phase
$66,500.00USD / Each
Labconco 6-Place Hooded Combo Kjeldahl Digestion/Distillation Apparatus, Single Phase; 230 VAC, 60 Hz EW-78909-12 Mfr # 2117612
COMPARE
800 6 230, 1 phase
$66,500.00USD / Each
Labconco 6-Place Hooded Combo Kjeldahl Digestion/Distillation Apparatus, Three Phase; 230 VAC, 60 Hz EW-78909-13 Mfr # 2117613
COMPARE
800 6 230, 3 phase
$66,500.00USD / Each
Showing 1- 7 of 7