Labconco Kjeldahl Digestion Apparatus
Ideal for laboratories that require a separate digestion station
  • Individual infinite control heat control knobs for precision heating
  • Accepts 500-mL or 800-mL Kjeldahl flasks
  • Curved heater cradles round-bottom flasks for great temperature uniformity
  • Powder-coated steel construction frame provide durability
The Kjeldahl digestion units are designed to help determine the total nitrogen content within materials such as feeds, grains, soils, fertilizers, plant tissue, water, organic wastes, and food products. Use the two- and six-place Kjeldahl digestion apparatus as a separate digestion station or as an addition to an existing Kjeldahl apparatus.
All models feature individual infinite-control knobs that provide variable heat input from 20 to 100% capacity for excellent heat control. Each 600-watt curved heater cradles Kjeldahl flasks for fast and evenly distributed heat. The digestion apparatus accepts 500- or 800-mL Kjeldahl flasks; simply move the heating element assembly closer or away from the manifold to accommodate different sizes. The chemical-resistant chlorinated polyvinyl chloride (CPVC) fume manifold with heat-resistant PTFE nipples draws chemical fumes from the digestion flasks for user safety. Select from two-place benchtop unit or six-place stand-alone units.
$7,695.00 - $23,345.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flask Size (mL) Number of Positions Power (VAC)
Availability Pricing
Labconco 6-Place Kjeldahl Digestion Apparatus, Single Phase; 230 VAC, 60 Hz EW-78909-14 Mfr # 2125612
COMPARE
800 6 230, 1 phase
$23,345.00USD / Each
Labconco 6-Place Kjeldahl Digestion Apparatus, Single Phase; 115 VAC, 60 Hz EW-78909-15 Mfr # 2125611
COMPARE
800 6 115, 1 phase
$23,345.00USD / Each
Labconco 6-Place Kjeldahl Digestion Apparatus, Three Phase; 230 VAC, 60 Hz EW-78909-16 Mfr # 2125613
COMPARE
800 6 230, 3 phase
$23,345.00USD / Each
Labconco 2-Place Kjeldahl Digestion Apparatus; 115 VAC, 50/60 Hz EW-78909-17 Mfr # 2128401
COMPARE
800 2 115
$7,695.00USD / Each