Labconco Protector® Echo™ Filtered Fume Hood
No ductwork – easily adapts to your changing lab spaces
  • Uses the Erlab® GFH filtration technology and Neutrodine® filters
  • Filtration allows you to work safely with a broader range of chemicals
  • Provides the space, utility and options of a traditional hood without potential challenges of existing infrastructure or lab configurations
More details and product features
Protector Echo hoods require no connection to ductwork — initial building infrastructure change costs are minimized and allow the hood to be placed in hard-to-duct areas such as the center or bottom level of multi-level buildings. The Protector Echo filtered fume hood features an Eco-Foil air foil with clean-sweep openings, fiberglass-reinforced panel liner, fully closing sash, and optional electrical receptacles and service fixtures. The Protector Echo hood creates significant energy savings because it doesn''t exhaust tempered room air from the lab and requires no make-up air! It also eliminates air pollutant emissions.
GreenFumeHood® (GFH) filtration technology allows the Protector Echo filtered fume hood to perform well beyond the SEFA 9-2010 DH III definition. User safety is maintained through numerous built-in features that constantly monitor and prevent unsafe conditions without relying on user modifications or practices.
The Neutrodine Filter has been tested and found efficient on over 500 chemicals, and may last two years or more before requiring replacement.
The hood is constructed of a glacier white powder-coated steel exterior and a chemical-resistant, fiberglass-reinforced, composite panel liner. The vertical-rising sash is a single piece of tempered safety glass with cable pulley, sash weight and powder-coated aluminum sash handle and tracks with Clean-Sweep™ airflow openings. Sash stop is at 16" sash opening height. Sash opens to 28" high for loading. The sightline from the work surface and header panel is 37.5" (95.3 cm) high.
Optional accessories include ADA remote control, gGuard® communication software, HEPA filter, and sash opening reduction wing kits. Contact an Application Specialist for details
The Protector Echo benchtop fume hood is also available with side windows or 360° windows, as well as 230 VAC power — contact an Application Specialist.
Note: In order to properly supply the right filtration for your application, the Lifetime Payback Questionnaire (LPQ) form is REQUIRED for all Protector Echo hood orders. It must be completed in full, submitted, reviewed, and approved before an order can be processed.
$22,665.00 - $31,620.00USD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Exterior Height (in) Exterior Width (in) Description
Availability Pricing
Labconco Echo 180410000 Filtered Fume Hood 4', 115V; No Service Fixtures or Outlets EW-32535-02 Mfr # 180410000
COMPARE
66.5 48 Filtered Fume Hood 4', 115V; No Service Fixtures or Outlets
$22,665.00USD / Each
Labconco Echo 180410001 Filtered Fume Hood 4', 115V; Two Service Fixtures EW-32535-04 Mfr # 180410001
COMPARE
66.5 48 Filtered Fume Hood 4', 115V; Two Service Fixtures
$23,565.00USD / Each
Labconco Echo 180410002 Filtered Fume Hood 4', 115V; Two Service Fixtures and Duplex Outlet EW-32535-06 Mfr # 180410002
COMPARE
66.5 48 Filtered Fume Hood 4', 115V; Two Service Fixtures and Duplex Outlet
$23,690.00USD / Each
Labconco Echo 180510000 Filtered Fume Hood 5', 115V; No Service Fixtures or Outlets EW-32535-08 Mfr # 180510000
COMPARE
66.5 60 Filtered Fume Hood 5', 115V; No Service Fixtures or Outlets
$24,370.00USD / Each
Labconco Echo 180510001 Filtered Fume Hood 5', 115V; Two Service Fixtures EW-32535-10 Mfr # 180510001
COMPARE
66.5 60 Filtered Fume Hood 5', 115V; Two Service Fixtures
$25,270.00USD / Each
Labconco Echo 180510002 Filtered Fume Hood 5', 115V; Two Service Fixtures and Duplex Outlet EW-32535-12 Mfr # 180510002
COMPARE
66.5 60 Filtered Fume Hood 5', 115V; Two Service Fixtures and Duplex Outlet
$25,390.00USD / Each
Labconco Echo 180610000 Filtered Fume Hood 6', 115V; No Service Fixtures or Outlets EW-32535-14 Mfr # 180610000
COMPARE
66.5 72 Filtered Fume Hood 6', 115V; No Service Fixtures or Outlets
$26,060.00USD / Each
Labconco Echo 180610001 Filtered Fume Hood 6', 115V; Two Service Fixtures EW-32535-16 Mfr # 180610001
COMPARE
66.5 72 Filtered Fume Hood 6', 115V; Two Service Fixtures
$26,970.00USD / Each
Labconco Echo 180610002 Filtered Fume Hood 6', 115V; Two Service Fixtures and Duplex Outlet EW-32535-18 Mfr # 180610002
COMPARE
66.5 72 Filtered Fume Hood 6', 115V; Two Service Fixtures and Duplex Outlet
$27,085.00USD / Each
Labconco Echo 180810000 Filtered Fume Hood 8', 115V; No Service Fixtures or Outlets EW-32535-20 Mfr # 180810000
COMPARE
66.5 96 Filtered Fume Hood 8', 115V; No Service Fixtures or Outlets
$30,595.00USD / Each
Labconco Echo 180810001 Filtered Fume Hood 8', 115V; Two Service Fixtures EW-32535-22 Mfr # 180810001
COMPARE
66.5 96 Filtered Fume Hood 8', 115V; Two Service Fixtures
$31,505.00USD / Each
Labconco Echo 180810002 Filtered Fume Hood 8', 115V; Two Service Fixtures and Two Duplex Outlet EW-32535-24 Mfr # 180810002
COMPARE
66.5 96 Filtered Fume Hood 8', 115V; Two Service Fixtures and Two Duplex Outlet
$31,620.00USD / Each
Showing 1- 12 of 12