Labconco Purifier Axiom Class II Type C1 Biosafety Cabinet
Operate in recirculated filtration or exhaust mode while maintaining Class II safety
  • The world''s first Class II Biosafety Cabinet that simplifies selection
  • Improved flexibility and chemical safety over both Type A and Type B biosafety cabinets
  • Provides lifetime operational cost savings over Type B biosafety cabinets
The Purifier Axiom Class II, Type C1 is the first biological safety cabinet that can be used in a recirculating Type A-mode for standard microbiological work, or can be connected to an exhaust system to function in Type-B mode for handling hazardous chemical vapors or radionuclides. Because of the efficient, well-defined airflow pattern over its work surface, the Axiom is safer and easier to operate than traditional biosafety cabinets.
The Axiom provides protection for personnel, products, and the laboratory environment by filtering hazardous particulates and aerosols, such as agents that require Biosafety Level 1, 2 or 3 containment, with the option of venting away hazardous fumes. Type C1 enclosures may also be used for a wide variety of applications that incorporate antineoplastic drugs, genetic material, carcinogens, allergens, or other airborne hazards.
The exclusive design of the Chem-Zone™ maximizes chemical safety by creating a large, clearly defined area within the enclosure, bringing peace of mind during chemical handling. Other features that make the Axiom safe and easy to operate include its ergonomic Inclination™ Technology and the intuitive MyLogic™ Operating System.
The Axiom can be run as a Type A or Type B BSC (with or without ducting). A base stand is required (not included with all models). Models available with or without options (including UV light, 1/4-turn ball valve, Vacu-Pass cord/cable port) or base stand. The 10" sash height can be adjusted to 8" for additional personal protection.
$18,900.00 - $24,350.00USD / Each
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Exhaust Volume (CFM) Exterior Width (in) Description
Availability Pricing
Labconco Purifier 304410000 Biosafety Cabinet, 4', 115V EW-32521-00 Mfr # 304410000
COMPARE
387 to 480 54.3125 Biosafety Cabinet, 4', 115V
$18,900.00USD / Each
Labconco Purifier 304411000 Biosafety Cabinet, UV Lamp with Timer, 4', 115V EW-32521-02 Mfr # 304411000
COMPARE
387 to 480 54.3125 Biosafety Cabinet, UV Lamp with Timer, 4', 115V
$19,800.00USD / Each
Labconco Purifier 304410100 Biosafety Cabinet, w/ Telescoping Base, 4', 115V EW-32521-04 Mfr # 304410100
COMPARE
387 to 480 54.3125 Biosafety Cabinet, w/ Telescoping Base, 4', 115V
$19,800.00USD / Each
Labconco Purifier 304411100 Biosafety Cabinet, 4', 115V EW-32521-06 Mfr # 304411100
COMPARE
387 to 480 54.3125 Biosafety Cabinet, 4', 115V
$20,650.00USD / Each
Labconco Purifier 304410010 Biosafety Cabinet, 4', 230V EW-32521-08 Mfr # 304410010
COMPARE
387 to 480 54.3125 Biosafety Cabinet, 4', 230V
$18,900.00USD / Each
Labconco Purifier 304411010 Biosafety Cabinet, UV Lamp with Timer, 4', 230V EW-32521-10 Mfr # 304411010
COMPARE
387 to 480 54.3125 Biosafety Cabinet, UV Lamp with Timer, 4', 230V
$19,800.00USD / Each
Labconco Purifier 304410110 Biosafety Cabinet; w/ Telescoping Base, 4', 230V EW-32521-12 Mfr # 304410110
COMPARE
387 to 480 54.3125 Biosafety Cabinet; w/ Telescoping Base, 4', 230V
$19,800.00USD / Each
Labconco Purifier 304411110 Biosafety Cabinet, 4', 230V EW-32521-14 Mfr # 304411110
COMPARE
387 to 480 54.3125 Biosafety Cabinet, 4', 230V
$20,650.00USD / Each
Labconco Purifier 304610000 Biosafety Cabinet, 6', 115V EW-32521-16 Mfr # 304610000
COMPARE
556 to 684 78.3125 Biosafety Cabinet, 6', 115V
$22,550.00USD / Each
Labconco Purifier 304611000 Biosafety Cabinet, UV Lamp with Timer, 6', 115V EW-32521-18 Mfr # 304611000
COMPARE
556 to 684 78.3125 Biosafety Cabinet, UV Lamp with Timer, 6', 115V
$23,450.00USD / Each
Labconco Purifier 304610100 Biosafety Cabinet, w/ Telescoping Base, 6', 115V EW-32521-20 Mfr # 304610100
COMPARE
556 to 684 78.3125 Biosafety Cabinet, w/ Telescoping Base, 6', 115V
$23,500.00USD / Each
Labconco Purifier 304611100 Biosafety Cabinet, 6', 115V EW-32521-22 Mfr # 304611100
COMPARE
556 to 684 78.3125 Biosafety Cabinet, 6', 115V
$24,350.00USD / Each
Labconco Purifier 304610010 Biosafety Cabinet, 6', 230V EW-32521-24 Mfr # 304610010
COMPARE
556 to 684 78.3125 Biosafety Cabinet, 6', 230V
$22,550.00USD / Each
Labconco Purifier 304611010 Biological Safety Cabinet, 6-ft, UV lamp with timer, 230V EW-32521-26 Mfr # 304611010
COMPARE
556 to 684 78.3125 Biological Safety Cabinet, 6-ft, UV lamp with timer, 230V
$23,450.00USD / Each
Labconco Purifier 304610110 Biosafety Cabinet, w/ Telescoping Base, 6', 230V EW-32521-28 Mfr # 304610110
COMPARE
556 to 684 78.3125 Biosafety Cabinet, w/ Telescoping Base, 6', 230V
$23,500.00USD / Each
Labconco Purifier 304611110 Biosafety Cabinet, 6', 230V EW-32521-30 Mfr # 304611110
COMPARE
556 to 684 78.3125 Biosafety Cabinet, 6', 230V
$24,350.00USD / Each
Showing 1- 16 of 16
Need Help with this product?
Call Us At1-847-549-7600
Live chat