Labconco Telescoping Base Stands for Purifier Horizontal Clean Benches
Labconco Telescoping Base Stands support Purifier Horizontal Clean Benches
  • 1000 pound load capacity
Stands feature telescoping legs so you can select stand height during installation. Height is adjustable to eight positions in 1” increments from 27.5” to 34.5”. Frame is durable 1.75” tubular, powder-coated steel with rear crossbar supports. ADA compliant. Fixed leveling feet. 1000 lb (454 kg) load capacity.
$929.00 - $1,242.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Exterior Depth (in) Exterior Width (in) Power (VAC)
Availability Pricing
Labconco 3613200 Telescoping Base Stand; 3' Purifier Horizontal Clean Bench EW-09116-74 Mfr # 3613200
COMPARE
29 35
$929.00USD / Each
Labconco 3888700 Telescoping Base Stand; 4' Purifier Horizontal Clean Bench EW-09116-76 Mfr # 3888700
COMPARE
29 47
$1,031.50USD / Each
Labconco 3857700 Telescoping Base Stand; 6' Purifier Horizontal Clean Bench EW-09116-78 Mfr # 3857700
COMPARE
29 71
$1,139.50USD / Each
Labconco 3857300 Telescoping Base Stand; 8' Purifier Horizontal Clean Bench EW-09116-80 Mfr # 3857300
COMPARE
29 96
$1,242.00USD / Each