Labnet Compact PCR Plate Centrifuge/Spinner
Quickly and efficiently improve your PCR yield
  • Accommodates two PCR plates
  • Spins down drops and condensation into sample
This unique centrifuge is less than 25% of the size of a standard plate centrifuge and weighs just 6 pounds. Unit accepts two skirted, non-skirted, or standard PCR plates in a vertical orientation to maximize your lab space. Easy operation — just load plates and push button to operate. When used before and after thermal cycling, yields are increased.
$725.00 - $761.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Capacity (mL) Max Speed (rpm) Power (VAC)
Availability Pricing
Labnet MPS 1000 Compact PCR / Microtiter Plate Centrifuge, 2500 rpm, 120VAC EW-17416-00 Mfr # C1000
COMPARE
2500 120
$725.00USD / Each
Labnet MPS 1000 Compact PCR / Microtiter Plate Centrifuge, 2500 rpm, 220VAC EW-17416-05 Mfr # C1000-230V
COMPARE
2500 230
$761.00USD / Each