Labnet Prism™ Air-Cooled and Refrigerated Microcentrifuges

Powerful motor with a small footprint
  • Deliver super fast deceleration and acceleration with the powerful brushless motor
  • Unique imbalance detection system with auto shut-off
These centrifuges deliver speed and high performance with the super fast motor. Original motor isolation system eliminates vibrations even at fast speeds and will automatically shut off if imbalance is detected. Easy-access rotor allows for quick loading, and adapter options provide versatility for your needs. Solidaluminum rotor can withstand repeated autoclave cycles. Large LCD shows actual spread in rpm or rcf, and time.
$2,390.00 - $5,600.00USD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Min Temperature (° C) Power (VAC)
Availability Pricing
Labnet C2500 Microcentrifuge; 120 VAC/60 Hz EW-17707-01 Mfr # C2500
COMPARE
Ambient 120
$2,510.00USD / Each
Labnet C2500-230V Microcentrifuge; 230 VAC/50 Hz EW-17707-03 Mfr # C2500-230V
COMPARE
Ambient 220
$2,390.00USD / Each
Labnet Prism R Refrigerated Microcentrifuge; 120 VAC/60 Hz EW-17707-07 Mfr # C2500-R
COMPARE
-10 120
$5,600.00USD / Each
Labnet Prism R Refrigerated Microcentrifuge; 230 VAC/50 Hz EW-17707-09 Mfr # C2500-R-230V
COMPARE
-10 220
$5,600.00USD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat