Labnet Spectrafuge™ 24D Digital Microcentrifuge
Precise digital control, high capacity, and a maintenance-free brushless drive
  • Exceptionally quite and cool running
  • Large display of rpm or g-force
  • Unique 24x1.5/2.0mL easy-access rotor
  • Pick your favorite color!
The digital microprocessor precisely regulates all aspects of operation. Run times from 30 seconds to 30 minutes, or continuously are easily set with the timer knob. Press and hold the knob for a quick spin — elapsed time is shown on the display.
$1,812.00 - $2,112.00USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Housing Color Power (VAC)
Availability Pricing
Labnet C2400-T-230V Microcentrifuge, Teal; 230 VAC/50 Hz EW-17410-45 Mfr # C2400-T-230V
COMPARE
Teal 220
$2,112.00USD / Each
Labnet C2400 Microcentrifuge, Grey; 120 VAC/60 Hz EW-17415-00 Mfr # C2400
COMPARE
Gray 120
$1,812.00USD / Each
Labnet C2400-B MicroCentrifuge, Blue; 120 VAC/60Hz EW-17415-10 Mfr # C2400-B
COMPARE
Blue 120
$1,938.00USD / Each
Labnet C2400-B-230V MicroCentrifuge, Blue; 230 VAC/50Hz EW-17415-15 Mfr # C2400-B-230V
COMPARE
Blue 220
$1,812.00USD / Each
Labnet C2400-P MicroCentrifuge, Purple; 120 VAC/60Hz EW-17415-20 Mfr # C2400-P
COMPARE
Purple 120
$1,938.00USD / Each
Labnet C2400-P-230V MicroCentrifuge, Purple; 230 VAC/50Hz EW-17415-25 Mfr # C2400-P-230V
COMPARE
Purple 220
$1,938.00USD / Each
Labnet C2400-R MicroCentrifuge, Red; 120 VAC/60Hz EW-17415-30 Mfr # C2400-R
COMPARE
Red 120
$1,812.00USD / Each
Labnet C2400-T MicroCentrifuge, Teal; 120 VAC/60Hz EW-17415-40 Mfr # C2400-T
COMPARE
Teal 120
$1,836.00USD / Each
Showing 1- 8 of 8