Ladder Design Cable Ties
Increased flexibility.
  • Self storing stub.
These ties are injection-molded of 6/6 nylon— won’t corrode or stretch. Durable locking mechanism holds securely even when used to bundle vibrating or flexing materials.
Ties continue to hold tension even in oily spray or wet environments. Ties are self-extinguishing as per UL 94 Flam. class: V-2. Ties withstand temperatures to 185degF (85degC). All ties remain flexible to –40degF (–40degC). Efficient range of ties may be higher or lower in your specific application. Melting temperature is 495degF (257degC).
$65.00 - $86.00USD / Bag of 1000
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Bundle Diameter (in) Length (in) Min Loop Strength (lbs)
Availability Pricing
COMPARE
1 3/4 7 40
$65.00USD / Bag of 1000
COMPARE
3 11 40
$86.00USD / Bag of 1000
Showing 1- 2 of 2
Need Help with this product?
Call Us At1-847-549-7600
Live chat