Lascar Electronics 21CFR-Compatible Temperature/RH/Dew Point Data Logger
Data encryption and audit tracking ensure compliance
  • All data and audit trails stored in an encrypted and unalterable format
  • Green and red LEDs flash to signal logging and alarm status
  • Configurable alarms, sampling rate, and start time customize logger for any application
  • IP67-rated housing protects logger from soil and moisture (cap must be fitted)
This temperature and humidity data logger provides data in 21CFR Part 11-compliant format for FDA-regulated industries. Configuration and data analysis software is available free of charge from the manufacturer. Software allows you to choose temperature units, sampling rate, start time, high and low alarm thresholds, and a unique ID for each logger. Security features include permission assignments for individual users, email alerts for failed login attempts, comments attachable to specific readings, and digital signatures. For convenience, the logger has an LCD screen which can cycle through current, minimum, and maximum readings for temperature and humidity. Flashing green and red LED indicators show logging status, alarm conditions, and low battery at a glance.
ITEM(S) ON SALE
$101.60 - $147.80USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Temperature Accuracy (° C) Description
Availability Pricing
Lascar Electronics EL-21CFR-2-LCD+ EasyLog 21CFR FDA-Compatible High Accuracy Temperature, Humidity, Dew Point Data Logger EW-18000-37 Mfr # EL-21CFR-2-LCD+
COMPARE
80 ±0.1° FDA-Compatible High Accuracy Temperature, Humidity, Dew Point Data Logger
$143.60USD / Each
$179.50USD / Each
Lascar Electronics EL-21CFR-TP-LCD+ EasyLog 21CFR FDA-Compatible High Accuracy Temperature Data Logger with Probe EW-18000-38 Mfr # EL-21CFR-TP-LCD+
COMPARE
125 ±0.45° FDA-Compatible High Accuracy Temperature Data Logger with Probe
$147.80USD / Each
$184.75USD / Each
Lascar Electronics EL-21CFR-2-LCD EasyLog 21CFR FDA-Compatible Temperature, Humidity, Dew Point Data Logger EW-18013-05 Mfr # EL-21CFR-2-LCD
COMPARE
80 ±0.55° from 5 to 60° FDA-Compatible Temperature, Humidity, Dew Point Data Logger
$120.80USD / Each
$151.00USD / Each
Lascar Electronics EL-21CFR-1-LCD EasyLog 21CFR FDA-Compatible Temperature Data Logger EW-18013-06 Mfr # EL-21CFR-1-LCD
COMPARE
80 ±1° FDA-Compatible Temperature Data Logger
$101.60USD / Each
$127.00USD / Each
Lascar Electronics EL-21CFR-TP-LCD EasyLog 21CFR FDA-Compatible Temperature Data Logger with Probe EW-18013-07 Mfr # EL-21CFR-TP-LCD
COMPARE
125 ±0.6° from -5 to 40° FDA-Compatible Temperature Data Logger with Probe
$126.20USD / Each
$157.75USD / Each