Lascar PanelPilot Smart Graphic Panel Meter Displays
Your fastest route to a personalized display!
  • Display connects to a PC via USB port for customization of color, units, logo, and graphical or numerical representation of measurements
  • Free PanelPilot software download is compatible with Windows® XP, 2000, Vista, and Windows 7
  • Panel mount with the included hardware, or leave free standing
These versatile color TFT graphic meter displays are user-programmable to show a variety of styles previously supported by traditional panel meters. Easily configure with the free PanelPilot software to display information through bar graph, analog, or numeric meter styles with user-defined color, text, units, and input scaling. A library of predefined elements such as meters, buttons, and switches is constantly being updated and expanded. USB to micro-USB cable is required for PC connection and programming. A standard null-modem cable will not power the unit as well as transfer data for programming, please order the 18000-58 interface cable (not included). Once programmed, the display can be left connected to a PC and powered through the USB port, or disconnected and powered through the 4 to 30 VDC terminal and mounted where needed. A USB to AC power adapter (part number 18000-59) is also available so you may plug the meter into a standard North American wall outlet.
$132.00 - $264.00USD / Each
7 variations of this product are available.
See All Product Options Below
RESOURCES
+ SAVE TO MY DOCS (PDFs)
7 PRODUCT OPTIONS
Compareitem specifications
Input Type Max DC Voltage VDC (VDC)
Availability Pricing
Lascar Electronics SGD 24-M PanelPilot 2.4" Graphic Panel Meter; 0-40 VDC Input EW-18000-17 Mfr # SGD 24-M
COMPARE
40
$132.00USD / Each
Lascar Electronics SGD 28-M PanelPilot 2.8" Graphic Panel Meter; 0-40 VDC Input EW-18000-18 Mfr # SGD 28-M
COMPARE
40
$150.00USD / Each
Lascar SGD 35-M PanelPilot 3.5" Graphic Panel Meter; 0-40 VDC Input EW-18000-19 Mfr # SGD 35-M
COMPARE
40
$175.00USD / Each
Lascar SGD 24-M420 PanelPilot 2.4" Graphic Panel Meter; 4-20 mA Input EW-18000-20 Mfr # SGD 24-M420
COMPARE
$152.00USD / Each
Lascar SGD 28-M421 PanelPilot 2.8" Graphic Panel Meter; 4-20 mA Input EW-18000-21 Mfr # SGD 28-M420
COMPARE
$179.00USD / Each
Lascar SGD 35-M422 PanelPilot 3.5" Graphic Panel Meter; 4-20 mA Input EW-18000-22 Mfr # SGD 35-M420
COMPARE
$214.00USD / Each
Lascar SGD 24-M-IP PanelPilot 2.4" Waterproof Graphic Panel Meter; 0-40 VDC In EW-18000-23 Mfr # SGD 24-M-IP
COMPARE
40
$264.00USD / Each
Showing 1- 7 of 7
Need Help with this product?
Call Us At1-847-549-7600
Live chat