Lascar Electronics Wireless Temperature and Temperature/Humidity Data Loggers with Optional Cloud Storage
Wirelessly stream temperature and humidity data through Wi-Fi
  • Communicate with PC or EasyLog Cloud using any Wi-Fi router
  • Get real-time data anywhere using free, downloadable EasyLog software
  • Change sensor settings remotely and get email alerts when alarm triggers
  • Stores sensor data in memory if temporarily disconnected from Wi-Fi
  • Configure high and low alarms
The EL-WiFi data logging sensors wirelessly stream environmental data at regular intervals to a host PC. Position sensor using built-in Wi-Fi range detection indicator; once set up within range of a Wi-Fi router, the sensor stays where you want it and you can monitor the data from your PC. The EL-WiFi sensors are 802.11b compliant and support security protocols WEP, WPA/WPA2-PSK—unlike many other wireless data loggers, these sensors communicate through any Wi-Fi router available worldwide. Use the LCD to read data locally, including min/max readings, Wi-Fi connection and signal strength. Data loggers are IP55-rated and include rechargeable batteries.
ITEM(S) ON SALE
$131.75 - $315.40USD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Input Type Max Temperature (° F) Temperature Accuracy (° C)
Availability Pricing
Lascar Electronics EL-WIFI-T+ High Accuracy Wireless Temperature Data Logger EW-18000-08 Mfr # EL-WIFI-T+
COMPARE
Temperature 140 ±0.1 (10 to 50), ±0.15 (all other ranges)
$143.60USD / Each
$179.50USD / Each
Lascar Electronics EL-WIFI-TH+ High Accuracy Wireless Temperature/RH Data Logger EW-18000-09 Mfr # EL-WIFI-TH+
COMPARE
Temperature and humidity 140 ±0.2 (5 to 60), ±0.8 (all other ranges)
$239.25USD / Each
Lascar Electronics EL-WIFI-TP+ High Accuracy Wireless Temperature Data Logger w/ Probe EW-18000-11 Mfr # EL-WIFI-TP+
COMPARE
Temperature (one external) 257 ±0.2 (-15 to 80), ±0.6 (all other ranges)
$220.20USD / Each
$275.25USD / Each
Lascar Electronics EL-Wi-Fi-TC Wireless Thermocouple Temperature Data Logger EW-18000-12 Mfr # EL-WIFI-TC
COMPARE
Temperature (J, K, N, or T thermocouple) 2372 ±1.5
$191.40USD / Each
$239.25USD / Each
Lascar Electronics EL-WiFi-TH Wireless Temperature & Humidity Data Logging Sensor EW-23039-01 Mfr # EL-WIFI-TH
COMPARE
Temperature and humidity 140
$153.60USD / Each
$192.00USD / Each
Lascar Electronics EL-WiFi-TP Wireless Temperature Datalogger with Probe EW-23039-02 Mfr # EL-WIFI-TP
COMPARE
Temperature (one external) 257 ±1.5
$153.60USD / Each
$192.00USD / Each
Lascar Electronics EL-WiFi-T Wireless Temperature Datalogger EW-23039-03 Mfr # EL-WIFI-T
COMPARE
Temperature 140 ±0.3 (5 to 60), ±0.8 (all other ranges)
$131.75USD / Each
Lascar Electronics EL-WIFI-DTC Dual-Channel Thermocouple Probe Temperature Data Logger EW-23039-07 Mfr # EL-WIFI-DTC
COMPARE
Temperature (J, K, N, or T thermocouple) 2372 ±1.5
$315.40USD / Each
$394.25USD / Each
Lascar Electronics EL-WIFI-DTP+ Dual-Channel Thermistor Probe Temperature Data Logger EW-23039-08 Mfr # EL-WIFI-DTP+
COMPARE
Temperature (dual-channel thermistor) 257 ±0.2 (-15 to 80), ±0.6 (all other ranges)
$310.40USD / Each
$388.00USD / Each
Showing 1- 9 of 9