Leeson NEMA Type 56C-Face DC Motors
Low-cost DC motors available in numerous configurations to meet your needs
  • Power pulleys, belts, pumps, or other machinery
  • Operate low-voltage models (12 or 24 VDC) with battery or solar power for fixed-speed applications
  • Explosion-proof models are Class 1, Groups C & D; Class II, Groups F & G rated
DC motors are suited for a variety of environments.
Totally enclosed, fan-cooled (TEFC) motors utilize an external fan to circulate outside air to the motor for applications where the motor will be running continuously.
Totally enclosed, nonvented (TENV) motors are designed without an air opening to protect against dust and other debris.
Explosion-proof (XPRF) motors are approved for areas where hazardous materials are used.
Washdown (WDN) motors are suitable for applications requiring regular washing or sanitizing cycles.
To vary the speed on a DC motor, order a DC controller at the side.
$710.00 - $2,540.00USD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Max Motor RPM (rpm) Motor Frame Size Motor hp
Availability Pricing
Leeson 098004 NEMA Type 56C-face TEFC motor, 1/3 hp, 1750 rpm, 90 VDC EW-70071-00 Mfr # 098004
COMPARE
1750 56C 1/3
$719.00USD / Each
Leeson 098006 NEMA Type 56C-face TEFC motor, 1/2 hp, 2500 rpm, 90 VDC EW-70071-10 Mfr # 098006
COMPARE
2500 56C 1/2
$775.00USD / Each
Leeson 098000 NEMA Type 56C-face TEFC motor, 1/2 hp, 1750 rpm, 90 VDC EW-70071-20 Mfr # 098000
COMPARE
1750 56C 1/2
$785.00USD / Each
Leeson 098008 NEMA Type 56C-face TEFC motor, 1/2 hp, 1750 rpm, 180 VDC EW-70071-30 Mfr # 098008
COMPARE
1750 56C 1/2
$781.00USD / Each
Leeson 108022 NEMA Type 56C-face TEFC motor, 1 hp, 1750 rpm, 90 VDC EW-70071-40 Mfr # 108022
COMPARE
1750 56C 1
$1,370.00USD / Each
Leeson 108023 NEMA Type 56C-face TEFC motor, 1 hp, 1750 rpm, 180 VDC EW-70071-50 Mfr # 108023
COMPARE
1750 56C 1
$1,560.00USD / Each
Leeson 108045 NEMA Type 56C-face TENV motor, 1/4 hp, 1800 rpm, 12 VDC EW-70072-00 Mfr # 108045
COMPARE
1800 56C 1/4
$710.00USD / Each
Leeson 108046 NEMA Type 56C-face TENV motor, 1/3 hp, 1800 rpm, 12 VDC EW-70072-10 Mfr # 108046
COMPARE
1800 56C 1/3
$727.00USD / Each
Leeson 108050 NEMA Type 56C-face TENV motor, 1/3 hp, 1800 rpm, 24 VDC EW-70072-20 Mfr # 108050
COMPARE
1800 56C 1/3
$767.00USD / Each
Leeson 108423 NEMA Type 56C-face WDN motor, 1/4 hp, 1750 rpm, 90 VDC EW-70073-00 Mfr # 108423
COMPARE
1750 56C 1/4
$895.00USD / Each
Leeson 118015 NEMA Type 56C-face XPRF motor, 1/3 hp, 1750 rpm, 90 VDC EW-70074-00 Mfr # 118015
COMPARE
1750 56C 1/3
$2,540.00USD / Each
Leeson 118016 NEMA Type 56C-face XPRF motor, 1/2 hp, 1750 rpm, 90 VDC EW-70074-10 Mfr # 118016
COMPARE
1750 56C 1/2
$2,350.00USD / Each
Showing 1- 12 of 12
Need Help with this product?
Call Us At1-847-549-7600
Live chat