Hand-Operated Lever Drum Pumps
Metal-body pumps for use in tight work areas where drums containing flammables are stored
  • Curved discharge spout for convenient hose extension
  • Designed for use in industrial, agricultural, and automotive industries
  • Model 06511-00 is ideal for use with high-viscosity fluids
  • Materials vary from Stainless steel to steel bodies with PTFE or Delrin acetal construction allow for easy handling of the toughest fluids
Lever drum pump model 06511-05 is able to dispense kerosene, antifreeze, lightweight oils, non-water-based and non-corrosive detergents, waxes, or disinfectants. High viscosity model 06511-00 can be used with paraffin, motor oil, cutting oil and other viscous lubricants. This pump comes with a high-angle discharge spout with screen and a curved discharge spout.
$57.10 - $342.50USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Tube Length, Inlet (in) Wetted Materials
Availability Pricing
COMPARE
140 32 Steel pump body, Delrin® acetal piston, neoprene seals
$89.80USD / Each
COMPARE
140 36 Zinc-plated pump body, steel piston, neoprene seals
$57.10USD / Each
COMPARE
140 40 Stainless Steel, PTFE
$342.50USD / Each
COMPARE
40 Stainless Steel, PTFE
$129.00USD / Each