LEVIFLOW® Clamp-On Ultrasonic Flow Sensors
No moving parts—no particle generation—no contamination
  • Starter kit and flow sensor required to build a complete system
  • Use on your existing silicone or C-Flex tubing
  • Obtain flow measurement without intrusion
  • Reusable flow sensors—choose from 4-, 20-, or 80-LPM models
These flow systems are ideal for sterile applications. Flow sensors fit over your existing silicone or C-Flex tubing lines and are easily reusable—remove sensor clamp, dispose of flexible tubing, and fix sensor over a new line.
These non-invasive flow measurement systems have no moving parts to contaminate or generate particulates, and are accurate to 3%.
$2,300.50 - $3,075.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (LPM) Min Flow Rate (LPM) Process Connection
Availability Pricing
LeviFlow LFSC-08D-001 Clamp-On Ultrasonic Flow Sensor, 4 L/min EW-24500-18 Mfr # 100-30396
COMPARE
4 0 8 mm OD tubing
$2,300.50USD / Each
LeviFlow LFSC-12D-007 Clamp-On Ultrasonic Flow Sensor, 20 L/min EW-24500-19 Mfr # 100-30390
COMPARE
20 0 12 mm OD tubing
$2,300.50USD / Each
LeviFlow LFSC-22D-005 Clamp-On Ultrasonic Flow Sensor, 80 L/min EW-24500-20 Mfr # 100-30391
COMPARE
80 0 22 mm OD tubing
$3,075.00USD / Each