LEVIFLOW® Single-Use Precision Ultrasonic Flowmeters
Obtain flow measurements of high-purity fluids
  • Order the starter kit and a flow sensor to build a complete system
  • Choose from sterile or non-sterile flow sensors covering a range of 80 mL/min to 80 LPM
  • Avoid contamination— systems don't contain moving or obtrusive parts
  • Sensors feature integrated sanitary clamp fittings
  • Easily configure flow sensor parameters via software
These ultrasonic flowmeter systems are designed for use in applications were cleanliness is required. Readings are accurate to 1%—high precision for high-purity fluids. Each system requires a starter kit and clamp-on flow sensor; order separately. The starter kit includes components to get a system in place, just choose a flow sensor at the desired flow range and sterility.
$312.20 - $428.90USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (LPM) Min Flow Rate (LPM) Wetted Materials
Availability Pricing
LeviFlow LFS-03SU-Z Single-Use Precision Ultrasonic Flow Sensor, 800 mL/min EW-24500-08 Mfr # 100-30375
COMPARE
0.8 0.035 Polypropylene
$372.60USD / Each
LeviFlow LFS-03SU-Z-G25 Single-Use Precision Ultrasonic Flow Sensor, 800 mL/min, gamma radiated EW-24500-09 Mfr # 100-30399
COMPARE
0.8 0.035 Polypropylene, gamma irradiated
$397.50USD / Each
LeviFlow LFS-06SU-Z Single-Use Precision Ultrasonic Flow Sensor, 8 L/min EW-24500-10 Mfr # 100-30377
COMPARE
8 1.7 Polypropylene
$312.20USD / Each
LeviFlow LFS-06SU-Z-G25 Single-Use Precision Ultrasonic Flow Sensor, 8 L/min, gamma radiated EW-24500-11 Mfr # 100-30400
COMPARE
8 1.7 Polypropylene, gamma irradiated
$339.30USD / Each
LeviFlow LFS-06SU-Z-SC1 Single-Use Precision Ultrasonic Flow Sensor, 8000 mL/min EW-24500-12 Mfr # 100-30394
COMPARE
8 0.08 Polypropylene
$402.20USD / Each
LeviFlow LFS-06SU-Z-SC1-G25 Single-Use Precision Ultrasonic Flow Sensor, 8000 mL/min, gamma radiated EW-24500-13 Mfr # 100-30406
COMPARE
8 0.08 Polypropylene, gamma irradiated
$428.90USD / Each
LeviFlow LFS-10SU-Z Single-Use Precision Ultrasonic Flow Sensor, 20 L/min EW-24500-14 Mfr # 100-30397
COMPARE
20 4.7 Polypropylene
$332.70USD / Each
LeviFlow LFS-10SU-Z-G25 Single-Use Precision Ultrasonic Flow Sensor, 20 L/min, gamma radiated EW-24500-15 Mfr # 100-30405
COMPARE
20 4.7 Polypropylene, gamma irradiated
$352.10USD / Each
LeviFlow LFS-20SU-Z Single-Use Precision Ultrasonic Flow Sensor, 80 L/min EW-24500-16 Mfr # 100-30379
COMPARE
80 18.8 Polypropylene
$380.10USD / Each
LeviFlow LFS-20SU-Z-G25 Single-Use Precision Ultrasonic Flow Sensor, 80 L/min, gamma radiated EW-24500-17 Mfr # 100-30404
COMPARE
80 18.8 Polypropylene, gamma irradiated
$409.60USD / Each
Showing 1- 10 of 10