Lightnin A-100 Two-Blade Folding Impellers
Two-blade folding impellers are perfect for mixing in vessels with narrow ports. Folded diameter is 11/16" for the 2" model and 1" for the 3" model. Impellers open fully when mixing.
$886.00 - $1,045.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Impeller Size (in) Impeller Size (mm) Material
Availability Pricing
Lightnin 800578 Two-Blade Folding Impeller, 5/16" Bore; 2" Dia EW-04543-00 Mfr # 800578
COMPARE
2 50.8 316 Stainless Steel
$886.00USD / Each
Lightnin 801146 Two-Blade Folding Impeller, 3/8" Bore; 3.0" Dia EW-04561-66 Mfr # 801146
COMPARE
3 76.2 316 Stainless Steel
$1,045.00USD / Each