LinkTech 20AC & 20PP Series Quick-Disconnect Fitting Body, Hose Barb without Valve
Simply press the stainless steel thumb latch to disconnect the fittings
  • Audible click lets you know the coupling is secure
  • Interchangeable with other similar industry thumb-latch fittings
Use these fittings to quickly connect and disconnect from your flow line. All fittings have 1/8" flow size, allowing you to mix and match between the fittings—simply order a body (female half) and an insert (male half). Couplings are compatible for use with gases and liquids. Choose between acetal for a wide operating temperature range and greater durability or polypropylene (PP) for stronger chemical resistance and ability to be gamma sterilized. Select from valved or straight-through (nonvalved) couplings—the valved fittings automatically seal the flow path when disconnected.
$82.00 - $184.00USD / Pkg of 25
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material
Availability Pricing
LinkTech 20AC Quick-Disconnect Body, acetal, 1/8" flow, nonvalved, 1/8" hose barb; 25/pk EW-16360-01 Mfr # 20AC-S2-02
COMPARE
1/8 Acetal
$82.00USD / Pkg of 25
LinkTech 20AC Quick-Disconnect Body, acetal, 1/8" flow, nonvalved, 1/4" hose barb; 25/pk EW-16360-02 Mfr # 20AC-S2-04
COMPARE
1/4 Acetal
$82.00USD / Pkg of 25
LinkTech 20AC Quick-Disconnect Body, acetal, 1/8" flow, nonvalved, 1/16" hose barb; 25/pk EW-16360-47 Mfr # 20AC-S2-01
COMPARE
1/16 Acetal
$141.00USD / Pkg of 25
LinkTech 20AC Quick-Disconnect Body, acetal, 1/8" flow, nonvalved, 3/16" hose barb; 25/pk EW-16360-48 Mfr # 20AC-S2-03
COMPARE
3/16 Acetal
$84.00USD / Pkg of 25
LinkTech 20PP Quick-Disconnect Body, almond PP, 1/8" flow, nonvalved, 1/8" hose barb; 25/pk EW-16361-01 Mfr # 20PP-S2-02MALD
COMPARE
1/8 Polypropylene
$184.00USD / Pkg of 25
LinkTech 20PP Quick-Disconnect Body, almond PP, 1/8" flow, nonvalved, 1/4" hose barb; 25/pk EW-16361-02 Mfr # 20PP-S2-04MALD
COMPARE
1/4 Polypropylene
$184.00USD / Pkg of 25
LinkTech 20PP Quick-Disconnect Body, almond PP, 1/8" flow, nonvalved, 1/16" hose barb; 25/pk EW-16361-31 Mfr # 20PP-S2-01MALD
COMPARE
1/16 Polypropylene
$184.00USD / Pkg of 25
Showing 1- 7 of 7