LinkTech 20AC and 20PP Series Quick-Disconnect Fitting Body, NPT(M) with Valve
Simply press the stainless steel thumb latch to disconnect the fittings
  • Audible click lets you know the coupling is secure
  • Interchangeable with other similar industry thumb-latch fittings
Use these fittings to quickly connect and disconnect from your flow line. All fittings have 1/8" flow size, allowing you to mix and match between the fittings—simply order a body (female half) and an insert (male half). Couplings are compatible for use with gases and liquids. Choose between acetal for a wide operating temperature range and greater durability or polypropylene (PP) for stronger chemical resistance and ability to be gamma sterilized. Select from valved or straight-through (nonvalved) couplings—the valved fittings automatically seal the flow path when disconnected.
$153.00 - $192.00USD / Pkg of 25
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Thread size NPT(M) (port 1) (in) Description
Availability Pricing
LinkTech 20AC Quick-Disconnect Body, acetal, 1/8" flow, valved, 1/8" NPT(M); 25/pk EW-16360-05 Mfr # 20ACV-SB1-02
COMPARE
Acetal 1/8 Quick-Disconnect Body, acetal, 1/8" flow, valved, 1/8" NPT(M), SS latch
$153.00USD / Pkg of 25
LinkTech 20AC Quick-Disconnect Body, acetal, 1/8" flow, valved, 1/4" NPT(M); 25/pk EW-16360-06 Mfr # 20ACV-SB1-04
COMPARE
Acetal 1/4 Quick-Disconnect Body, acetal, 1/8" flow, valved, 1/4" NPT(M), SS latch
$154.00USD / Pkg of 25
LinkTech 20PP Quick-Disconnect Body, almond PP, 1/8" flow, valved, 1/8" NPT(M); 25/pk EW-16361-05 Mfr # 20PPV-SE1-02MALD
COMPARE
Polypropylene 1/8 Quick-Disconnect Body, almond PP, 1/8" flow, panel mount, nonvalved, 1/4" hose barb, SS latch
$192.00USD / Pkg of 25
LinkTech 20PP Quick-Disconnect Body, almond PP, 1/8" flow, valved, 1/4" NPT(M); 25/pk EW-16361-06 Mfr # 20PPV-SE1-04MALD
COMPARE
Polypropylene 1/4 Quick-Disconnect Body, almond PP, 1/8" flow, valved, 1/8" NPT(M), SS latch
$192.00USD / Pkg of 25