LinkTech 20AC and 20PP Series Quick-Disconnect Fitting Insert, Hose Barb with Valve
Simply press the stainless steel thumb latch to disconnect the fittings
  • Audible click lets you know the coupling is secure
  • Interchangeable with other similar industry thumb-latch fittings
Use these fittings to quickly connect and disconnect from your flow line. All fittings have 1/8" flow size, allowing you to mix and match between the fittings—simply order a body (female half) and an insert (male half). Couplings are compatible for use with gases and liquids. Choose between acetal for a wide operating temperature range and greater durability or polypropylene (PP) for stronger chemical resistance and ability to be gamma sterilized. Select from valved or straight-through (nonvalved) couplings—the valved fittings automatically seal the flow path when disconnected.
$182.00 - $190.00USD / Pkg of 25
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Description
Availability Pricing
LinkTech 20AC Quick-Disconnect Insert, acetal, 1/8" flow, valved, 1/16" hose barb; 25/pk EW-16360-68 Mfr # 20ACV-PB2-01
COMPARE
1/16 Acetal Quick-Disconnect Insert, acetal, 1/8" flow, valved, 1/4" BSPT(M), SS latch
$190.00USD / Pkg of 25
LinkTech 20PP Quick-Disconnect Insert, almond PP, 1/8" flow, valved, 1/16" hose barb; 25/pk EW-16361-39 Mfr # 20PPV-PE2-01MALD
COMPARE
1/16 Polypropylene Quick-Disconnect Insert, almond PP, 1/8" flow, valved, 1/16" hose barb, SS latch
$182.00USD / Pkg of 25