LinkTech 20AC and 20PP Series Quick-Disconnect Fitting Insert, Hose Barb without Valve, Panel-Mount
Simply press the stainless steel thumb latch to disconnect the fittings
  • Audible click lets you know the coupling is secure
  • Interchangeable with other similar industry thumb-latch fittings
Use these fittings to quickly connect and disconnect from your flow line. All fittings have 1/8" flow size, allowing you to mix and match between the fittings—simply order a body (female half) and an insert (male half). Couplings are compatible for use with gases and liquids. Choose between acetal for a wide operating temperature range and greater durability or polypropylene (PP) for stronger chemical resistance and ability to be gamma sterilized. Select from valved or straight-through (nonvalved) couplings—the valved fittings automatically seal the flow path when disconnected.
$110.00 - $136.00USD / Pkg of 25
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Description
Availability Pricing
LinkTech 20AC Quick-Disconnect Insert, acetal, 1/8" flow, panel mount, nonvalved, 1/8" hose barb; 25/pk EW-16360-34 Mfr # 20AC-PB3-02
COMPARE
1/8 Quick-Disconnect Insert, acetal, 1/8" flow, panel mount, nonvalved, 1/8" hose barb, SS latch
$110.00USD / Pkg of 25
LinkTech 20AC Quick-Disconnect Insert, acetal, 1/8" flow, panel mount, nonvalved, 1/4" hose barb; 25/pk EW-16360-35 Mfr # 20AC-PB3-04
COMPARE
1/4 Quick-Disconnect Insert, acetal, 1/8" flow, panel mount, nonvalved, 1/4" hose barb, SS latch
$112.00USD / Pkg of 25
LinkTech 20AC Quick-Disconnect Insert, acetal, 1/8" flow, panel mount, nonvalved, 1/16" hose barb; 25/pk EW-16360-62 Mfr # 20AC-PB3-01
COMPARE
1/16 Quick-Disconnect Insert, acetal, 1/8" flow, nonvalved, 3/16" hose barb, SS latch
$136.00USD / Pkg of 25
LinkTech 20AC Quick-Disconnect Insert, acetal, 1/8" flow, panel mount, nonvalved, 3/16" hose barb; 25/pk EW-16360-63 Mfr # 20AC-PB3-03
COMPARE
3/16 Quick-Disconnect Insert, acetal, 1/8" flow, panel mount, nonvalved, 1/16" hose barb, SS latch
$110.00USD / Pkg of 25