LinkTech 20AC and 20PP Series Quick-Disconnect Fitting Insert, Hose Barb without Valve
Simply press the stainless steel thumb latch to disconnect the fittings
  • Audible click lets you know the coupling is secure
  • Interchangeable with other similar industry thumb-latch fittings
Use these fittings to quickly connect and disconnect from your flow line. All fittings have 1/8" flow size, allowing you to mix and match between the fittings—simply order a body (female half) and an insert (male half). Couplings are compatible for use with gases and liquids. Choose between acetal for a wide operating temperature range and greater durability or polypropylene (PP) for stronger chemical resistance and ability to be gamma sterilized. Select from valved or straight-through (nonvalved) couplings—the valved fittings automatically seal the flow path when disconnected.
$26.25 - $54.00USD / Pkg of 25
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Description
Availability Pricing
LinkTech 20AC Quick-Disconnect Insert, acetal, 1/8" flow, nonvalved, 1/8" hose barb; 25/pk EW-16360-14 Mfr # 20AC-PB2-02
COMPARE
1/8 Acetal Quick-Disconnect Insert, acetal, 1/8" flow, nonvalved, 1/8" hose barb, SS latch
$26.25USD / Pkg of 25
LinkTech 20AC Quick-Disconnect Insert, acetal, 1/8" flow, nonvalved, 1/4" hose barb; 25/pk EW-16360-15 Mfr # 20AC-PB2-04
COMPARE
1/4 Acetal Quick-Disconnect Insert, acetal, 1/8" flow, nonvalved, 1/4" hose barb, SS latch
$31.75USD / Pkg of 25
LinkTech 20AC Quick-Disconnect Insert, acetal, 1/8" flow, nonvalved, 1/16" hose barb; 25/pk EW-16360-60 Mfr # 20AC-PB2-01
COMPARE
1/16 Acetal Quick-Disconnect Insert, acetal, 1/8" flow, nonvalved, 1/4" BSPT(M), SS latch
$49.50USD / Pkg of 25
LinkTech 20AC Quick-Disconnect Insert, acetal, 1/8" flow, nonvalved, 3/16" hose barb; 25/pk EW-16360-61 Mfr # 20AC-PB2-03
COMPARE
3/16 Acetal Quick-Disconnect Insert, acetal, 1/8" flow, nonvalved, 1/16" hose barb, SS latch
$31.75USD / Pkg of 25
LinkTech 20PP Quick-Disconnect Insert, almond PP, 1/8" flow, nonvalved, 1/8" hose barb; 25/pk EW-16361-12 Mfr # 20PP-PE2-02MALD
COMPARE
1/8 Polypropylene Quick-Disconnect Insert, almond PP, 1/8" flow, nonvalved, 1/4" hose barb, SS latch
$51.50USD / Pkg of 25
LinkTech 20PP Quick-Disconnect Insert, almond PP, 1/8" flow, nonvalved, 1/4" hose barb; 25/pk EW-16361-13 Mfr # 20PP-PE2-04MALD
COMPARE
1/4 Polypropylene Quick-Disconnect Insert, almond PP, 1/8" flow, elbow, nonvalved, 1/8" hose barb, SS latch
$51.50USD / Pkg of 25
LinkTech 20PP Quick-Disconnect Insert, almond PP, 1/8" flow, nonvalved, 1/16" hose barb; 25/pk EW-16361-38 Mfr # 20PP-PE2-01MALD
COMPARE
1/16 Polypropylene Quick-Disconnect Insert, almond PP, 1/8" flow, nonvalved, 1/16" hose barb, SS latch
$54.00USD / Pkg of 25
Showing 1- 7 of 7