LinkTech 20AC and 20PP Series Quick-Disconnect Fitting Insert, NPT(M) with Valve
Simply press the stainless steel thumb latch to disconnect the fittings
  • Audible click lets you know the coupling is secure
  • Interchangeable with other similar industry thumb-latch fittings
Use these fittings to quickly connect and disconnect from your flow line. All fittings have 1/8" flow size, allowing you to mix and match between the fittings—simply order a body (female half) and an insert (male half). Couplings are compatible for use with gases and liquids. Choose between acetal for a wide operating temperature range and greater durability or polypropylene (PP) for stronger chemical resistance and ability to be gamma sterilized. Select from valved or straight-through (nonvalved) couplings—the valved fittings automatically seal the flow path when disconnected.
$156.00 - $201.00USD / Pkg of 25
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Thread size NPT(M) (port 1) (in) Description
Availability Pricing
LinkTech 20AC Quick-Disconnect Insert, acetal, 1/8" flow, valved, 1/8" NPT(M); 25/pk EW-16360-18 Mfr # 20ACV-PB1-02
COMPARE
Acetal 1/8 Quick-Disconnect Insert, acetal, 1/8" flow, valved, 1/8" NPT(M), SS latch
$156.00USD / Pkg of 25
LinkTech 20AC Quick-Disconnect Insert, acetal, 1/8" flow, valved, 1/4" NPT(M); 25/pk EW-16360-19 Mfr # 20ACV-PB1-04
COMPARE
Acetal 1/4 Quick-Disconnect Insert, acetal, 1/8" flow, valved, 1/4" NPT(M), SS latch
$161.00USD / Pkg of 25
LinkTech 20PP Quick-Disconnect Insert, almond PP, 1/8" flow, valved, 1/8" NPT(M); 25/pk EW-16361-16 Mfr # 20PPV-PE1-02MALD
COMPARE
Polypropylene 1/8 Quick-Disconnect Insert, almond PP, 1/8" flow, valved, 1/8" hose barb, SS latch
$201.00USD / Pkg of 25