LinkTech 20AC and PP Series Quick-Disconnect Fitting Body, BSPT(M) without Valve
Simply press the stainless steel thumb latch to disconnect the fittings
  • Audible click lets you know the coupling is secure
  • Interchangeable with other similar industry thumb-latch fittings
Use these fittings to quickly connect and disconnect from your flow line. All fittings have 1/8" flow size, allowing you to mix and match between the fittings—simply order a body (female half) and an insert (male half). Couplings are compatible for use with gases and liquids. Choose between acetal for a wide operating temperature range and greater durability or polypropylene (PP) for stronger chemical resistance and ability to be gamma sterilized. Select from valved or straight-through (nonvalved) couplings—the valved fittings automatically seal the flow path when disconnected.
$118.00 - $126.00USD / Pkg of 25
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Description
Availability Pricing
LinkTech 20AC Quick-Disconnect Body, acetal, 1/8" flow, nonvalved, 1/8" BSPT(M); 25/pk EW-16360-45 Mfr # 20AC-S1-02BSPT
COMPARE
Acetal Quick-Disconnect Body, acetal, 1/8" flow, nonvalved, 1/8" BSPT(M), SS latch
$118.00USD / Pkg of 25
LinkTech 20AC Quick-Disconnect Body, acetal, 1/8" flow, nonvalved, 1/4" BSPT(M); 25/pk EW-16360-46 Mfr # 20AC-S1-04BSPT
COMPARE
Acetal Quick-Disconnect Body, acetal, 1/8" flow, nonvalved, 1/4" BSPT(M), SS latch
$126.00USD / Pkg of 25