LinkTech 20CB Series Quick-Disconnect Fitting Body, NPT(M) with Valve
Chrome-plated brass construction withstands higher operating pressures
  • Audible click lets you know the coupling is secure
  • Buna N O-ring seal eliminates leaks
  • Interchangeable with other similar industry thumb-latch fittings
These food-grade chrome-plated brass fittings are well suited for food and beverage applications. The 1/8” flow size bodies and inserts are durable and can withstand operating pressures up to 250 psi (17.2 bar). Intuitive thumb-latch design makes disconnecting fittings quick and easy with one hand.
You can easily mix and match valved and straight-through fittings to suit your flow path needs—simply order a body (female half) and an insert (male half). Valved fittings automatically seal the flow path when disconnected.
$110.00USD / Pkg of 10
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Quick-Disconnect Flow Size (in) Thread size NPT(M) (port 1) (in)
Availability Pricing
LinkTech 20CB Quick-Disconnect Body, chrome brass, 1/8" flow, in-line, valved, 1/8" NPT(M); 10/pk EW-41404-05 Mfr # 20CBV-SB1-02
COMPARE
Chrome-plated brass 1/8 1/8
$110.00USD / Pkg of 10
LinkTech 20CB Quick-Disconnect Body, chrome brass, 1/8" flow, in-line, valved, 1/4" NPT(M); 10/pk EW-41404-06 Mfr # 20CBV-SB1-04
COMPARE
Chrome-plated brass 1/8 1/4
$110.00USD / Pkg of 10