LinkTech 30AC Series Quick-Disconnect Fittings Body, Hose Barb with Valve, Panel Mount
Extra reliability with less moving parts
  • Advanced plastic protection from mild chemicals in the work area
  • Audible click guarantees a secure connection with a thumb press
The advanced all-plastic fittings are durable, secure, and offer proven reliability with a long service life. Acetal material provides a wide operating temperature range and is resistant to most mild chemicals. The fittings feature fewer moving parts, an internal spring, and a plastic thumb latch designed to quickly connect and disconnect from your flow line.
All fittings have 1/8" flow size, allowing you to mix and match valved and straight-through (nonvalved) fittings to suit your flow path needs—simply order a body (female half) and an insert (male half). Valved fittings automatically seal the flow path when disconnected.
$166.00 - $205.00USD / Pkg of 25
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Length (mm) Quick-Disconnect Flow Size (in)
Availability Pricing
LinkTech 30AC Quick-Disconnect Body, acetal, 1/8" flow, panel mount, valved, 1/16" hose barb; 25/pk EW-41411-33 Mfr # 30ACV-SB3-01
COMPARE
1/16 36 1/8
$205.00USD / Pkg of 25
LinkTech 30AC Quick-Disconnect Body, acetal, 1/8" flow, panel mount, valved, 1/8" hose barb; 25/pk EW-41411-34 Mfr # 30ACV-SB3-02
COMPARE
1/8 42 1/8
$166.00USD / Pkg of 25
LinkTech 30AC Quick-Disconnect Body, acetal, 1/8" flow, panel mount, valved, 3/16" hose barb; 25/pk EW-41411-35 Mfr # 30ACV-SB3-03
COMPARE
3/16 46 1/8
$169.00USD / Pkg of 25
LinkTech 30AC Quick-Disconnect Body, acetal, 1/8" flow, panel mount, valved, 1/4" hose barb; 25/pk EW-41411-36 Mfr # 30ACV-SB3-04
COMPARE
1/4 46 1/8
$169.00USD / Pkg of 25